Whole Allergen
e213 Parrot feathers
e213 Parrot feathers Scientific Information
Type:
Whole Allergen
Display Name:
Parrot feathers
Allergen code:
e213
Family:
Psittacidae
Species:
Ara spp.
Route of Exposure:
Inhalation
Latin Name:
Ara spp.
Other Names:
Parrot
WHO/ICD-11 code:
XE69N
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Summary
Parrots are a group of colorful tropical bird species popular as pet birds. Most parrot species are found in tropical and sub-tropical forests across Central and South America, Australasia, Oceania, and Africa. Parrots are found to nest in holes and prefer to reside in wetter environments and warm regions. Some parrot blooms are found to be responsible for transporting soluble antigens to the lower airways and, therefore, can act as an adjuvant, enhancing the immunogenic reaction. Exposure to parrot feathers has been found to be associated with the bird-egg syndrome, asthma, allergic rhinitis, rhinoconjunctivitis, and hypersensitivity pneumonitis. Studies with parrot feather extract have reported the presence of few allergenic protein bands. Cross-reactivity is found between sera and feathers of bird species, including parrots, the canary, goose, hen, duck, and pigeon. Furthermore, budgerigar feather extract (containing albumin) is reported to be cross-reactive with egg yolk extract (containing Gal d 5).
Allergen
Nature
The Psittaciformes order includes 370 species and 576 subspecies as per the ITIS database (1). Parrots (belonging to the order Psittaciformes) are reported as a group of colorful tropical bird species, which exhibit distinctive feather colors (2) and are very popular pet birds (3). These birds possess a typical, well-developed upper beak (hooked rostrum) along with an adjacent featherless portion (cere). The upper beak in large parrots (e.g., macaws) is joined to the skull via a synovial joint. However, in smaller parrots (e.g., budgerigars), this joint is connected through an elastic zone (3).
Taxonomy
The birds belonging to the order Psittaciformes include parrots, macaws, cockatoos, and lories (3).
Taxonomic tree of Parrot feathers (4)
Domain Eukaryota
Kingdom Metazoa
Phylum Chordata
Subphylum Vertebtata
Class Aves
Order Psittaciformes
Family Psittacidae
Genus Ara
Species Ara spp.
Tissue
Allergenic proteins are reported to be present in the bird’s feather bloom, droppings, and skin scales. Some parrots, including parakeets, budgerigars, and cockatiels, produce large blooms, where keratin granules are reported to be responsible for transporting the soluble antigens to the lower airways and can act as an adjuvant, enhancing the immunogenic reaction (5).
Epidemiology
Worldwide distribution
In a study from an allergic population in Venezuela, serum specific-IgE (sIgE) to parrot feathers were observed in 24 out of 189 allergic patients with allergic rhinitis and/or asthma (6).
Occupational allergy to birds has been reported as one of the significant health issues among zoo bird keepers, which was addressed in a study involving Polish zoo employees (Swiderska-Kielbik, Krakowiak et al. 2011). The study assessed 200 bird zookeepers where allergen-sIgE against any bird allergen was found in 72 % of subjects, and sIgE against parrot feathers was detected among 25 % of the subjects (7)
Environmental Characteristics
Living environment
Parrots nest in holes and prefer to reside in wetter environments and warm regions (3, 8).
Worldwide distribution
Most parrot species are found in tropical and sub-tropical forests across Central and South America, Australasia and Oceania, and Africa (9).
Route of Exposure
Main
Inhalation of parrot feathers dust is considered the route of exposure that can trigger immunological reactions (5, 10).
Clinical Relevance
Allergy to parrots has been reported to be associated with rhinoconjunctivitis, asthma, allergic rhinitis, bird-egg syndrome, and bird fancier’s lung (11-15).
Asthma, allergic rhinitis, and rhinoconjunctivitis 
Bronchial asthma, allergic rhinitis, and rhinoconjunctivitis may result from exposure to budgerigar feathers, serum, or droppings (11, 15). 
Bird-egg syndrome
Bird-egg syndrome (typically based upon sensitization to bird allergens and allergic symptoms occurring after egg consumption) has been reported in adults due to sensitization to avian airborne allergens through feathers, sera, and droppings of pet birds, including parrots (12). 
Bird fancier’s lung 
Bird fancier’s lung is a common type of hypersensitivity pneumonitis triggered by frequent exposure to avian aeroantigens (including proteins present on the feathers and excreted in bird droppings) (13, 14).
In a study involving patients with bird fancier’s asthma, the highest sIgE antibody binding was observed among bird fanciers (the bird caregivers) who had contact with the feathers of the bird species, including parrots (10).
Prevention and Therapy
Prevention strategies
Avoidance
Completely avoiding the antigen source is considered the primary measure to reduce the amount of antigen in the respiratory system (12, 16). Additionally, patients are also suggested to thoroughly clean their house and install HEPA air cleaner (17).
Molecular Aspects
Allergenic molecules
To date (22-SEP-2021), no allergenic molecule has been identified from parrot feathers according to the WHO/IUIS (18).
However, a study has found well-defined major allergenic bands (through IgE immunoblots; 20-30 kDa and 67 kDa) with feather extracts and serum proteins of parrot, budgerigar, canary, pigeon, and hen (10).
Furthermore, another study found a 70 kDa allergenic protein band (based on IgE immunoblotting) with budgerigar (parrot) feather extract corresponding to the budgerigar serum albumin (12).
Cross-reactivity
Cross-reactivity was seen between sera and feathers of the tested bird species (including the parrot, canary, goose, hen, duck, and pigeon) in patients with bird fancier’s asthma (10).
Cross-reactivity was seen between egg yolk extract (Gal d 5; livetin) and budgerigar feather extract containing albumin (12).
Compiled By
Author:Turacoz Healthcare Solutions
Reviewer: Dr. Christian Fischer
Last reviewed: March  2022
References
  1. ITIS. Psittaciformes 2021 [18.11.2021]. Available from: https://www.itis.gov/servlet/SingleRpt/SingleRpt?search_topic=TSN&search_value=177404#null.
  2. Burtt EH, Jr., Schroeder MR, Smith LA, Sroka JE, McGraw KJ. Colourful parrot feathers resist bacterial degradation. Biol Lett. 2011;7(2):214-6.
  3. Harcourt-Brown N. Chapter 7. Psittacine Birds. .  Handbook of Avian Medicine  10.1016/B978-0-7020-2874-8.00007-9. ; 2009. p. 138-68.
  4. CABI. Ara ararauna 2019 [17-09-2021]. Available from: https://www.cabi.org/isc/datasheet/93352.
  5. Lopata AS, Magda. Evaluating allergenic responses to bird allergens. Current Allergy & Clinical Immunology 17 2004.
  6. Sanchez-Borges M, Capriles-Hulett A, Torres J, Ansotegui-Zubeldia IJ, Castillo A, Dhersy A, et al. Diagnosis of allergic sensitization in patients with allergic rhinitis and asthma in a tropical environment. Rev Alerg Mex. 2019;66(1):44-54.
  7. Swiderska-Kielbik S, Krakowiak A, Wiszniewska M, Nowakowska-Swirta E, Walusiak-Skorupa J, Sliwkiewicz K, et al. Occupational allergy to birds within the population of Polish bird keepers employed in zoo gardens. Int J Occup Med Environ Health. 2011;24(3):292-303.
  8. Carballo LD, Kaspar & Valcu, Mihai & Kempenaers, Bart. Body size and climate as predictors of plumage colouration and sexual dichromatism in parrots. Journal of Evolutionary Biology 33 101111/jeb13690 2020.
  9. Vergara-Tabares DL, Cordier JM, Landi MA, Olah G, Nori J. Global trends of habitat destruction and consequences for parrot conservation. Glob Chang Biol. 2020;26(8):4251-62.
  10. Tauer-Reich I, Fruhmann G, Czuppon AB, Baur X. Allergens causing bird fancier's asthma. Allergy. 1994;49(6):448-53.
  11. Gerth Van Wijk RVT, A. W. Dieges, P. H. Nasal allergy to avian antigens. Clin Allergy. 1987;17(6):515-21.
  12. Inomata N, Kawano K, Aihara M. Bird-egg syndrome induced by alpha-livetin sensitization in a budgerigar keeper: Successful induction of tolerance by avoiding exposure to avians. Allergol Int. 2019;68(2):282-4.
  13. Spagnolo P, Rossi G, Cavazza A, Bonifazi M, Paladini I, Bonella F, et al. Hypersensitivity Pneumonitis: A Comprehensive Review. J Investig Allergol Clin Immunol. 2015;25(4):237-50; quiz follow 50.
  14. Sullivan A, Shrestha P, Lanham T, Lanham E, Baba M. Bird Fancier's lung: An underdiagnosed etiology of dyspnea. Respir Med Case Rep. 2020;31:101288.
  15. van Toorenenbergen AWGvW, R. van Dooremalen, G. Dieges, P. H. Immunoglobulin E antibodies against budgerigar and canary feathers. Int Arch Allergy Appl Immunol. 1985;77(4):433-7.
  16. Caillaud D, Raobison R, Evrard B, Montcouquiol S, Horo K. [Domestic hypersensitivity pneumonitis]. Rev Mal Respir. 2012;29(8):971-7.
  17. ACAAI. Dust allergies 2018 [22-09-2021.]. Available from: https://acaai.org/allergies/allergic-conditions/dust-allergies/.
  18. WHO-IUIS. Parrot feather 2021 [22-09-2021]. Available from: http://www.allergen.org/search.php?allergensource=parrot+feather&searchsource=Search.
  19. Liu YN, Chen LA, Zhang ZY, Li QS. Parrot breeder's lung: first case report in China. Chin Med J (Engl). 1989;102(12):947-50.