Whole Allergen
e77 Budgerigar droppings
e77 Budgerigar droppings Scientific Information
Type:
Whole Allergen
Display Name:
Budgerigar droppings
Allergen code:
e77
Family:
Psittacidae
Latin Name:
Melopsittacus undulatus
WHO/ICD-11 code:
XE69N
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Summary
Direct or indirect contact with bird allergens frequently causes sensitization. Bird allergens may be major components in house dust.
Environment
Allergen Exposure
Geographical distribution
Budgerigar and Parakeet are the common names for a widespread group of small Parrots, native to the Indo-Malayan-Australian region and now the most popular cage birds in the world. Parakeets have long, pointed tails, unlike the chunky Lovebirds with which they are sometimes confused. The wild Budgerigar of Australia is usually green or blue with black and yellow markings; as a cage bird, however, it has been bred in many color varieties.
Environment
Large flocks, sometimes in the tens of thousands, inhabit the open grasslands in central Australia, nesting in the spring and summer in the southern areas of the continent. In most of the world, however, Budgerigars are familiar only as pets.
Allergens
No allergens from this bird have yet been fully characterised.
Canary and Budgerigar feathers contain IgE-binding antigens that are not present in the corresponding bird sera and droppings (1).
Potential Cross Reactivity
Cross-reactivity between Chicken and other phylogenetically related bird species may be expected, and in Chicken-allergic patients significant IgE titers to Parrot, Budgerigar, Chicken, Pigeon, Goose and Duck have been reported (2-3), even in patients without known exposure (6).
Clinical Experience
IgE-mediated reactions
Asthma, allergic rhinitis and allergic conjunctivitis may result on exposure to Budgerigars (1, 4).
In a prospective study, 258 adults were investigated for sensitisation against bird antigens (Budgerigar, Canary, Pigeon) using an intracutaneous test. Eighteen of 78 Budgerigar keepers reacted against Budgerigar feathers. Thirty (38%) showed a positive skin reaction with at least 1 of the 3 extracts tested. In non-bird-exposed persons a reaction was found in 18/96 (19%) against Budgerigar and in 24/96 (25%) against at least 1 of the 3 allergen extracts. Late reactions occurred in 5/70 (7%) Budgerigar keepers. The frequency of sensitisation against Budgerigar antigens increased to 48% of bird keepers (24/50) if the birds could fly around in their homes instead of being always caged (4/22, 18%). Three of the 78 Budgerigar keepers (4%) had a positive nasal provocation with feather extract, and they were skin-positive against all 3 bird antigens tested (6).
Extrinsic allergic alveolitis, also known as hypersensitivity pneumonitis, Bird Fancier's Lung and Farmer's Lung, is a disease of inflammation of the lung parenchyma in the terminal bronchioles and alveoli. Symptoms may start soon after exposure to bird allergens or after many years, and may include breathlessness, cough, occasional chills, and fever. Death may also result.
The disease occurs after exposure to organic dust, especially after close contact with Pigeons or other birds such as Budgerigars, Parrots, Canaries, Parakeets, Cockatiels, Doves or Finches. Exposure results in the development of immunoglobulin antibodies including IgE (6),  IgM (7),  IgA and various IgG subclasses (8-10. The antibodies may be found in the sera and saliva of patients (11) as well as in the sera of asymptomatic but exposed subjects (12). 
The allergenic proteins may be found in bird serum, droppings, skin scales, feathers and, in the case of Pigeons, Pigeon bloom (a waxy fine dust which coats the feathers of Pigeons). Contact may result from handling birds, cleaning their lofts, or exposure to the organic dust drifting down from a ceiling or roof where birds nest.
Diagnosis is based on a characteristic clinical picture and a typical x-ray pattern, accompanied by the presence of specific IgG antibodies (13).
The measurement of specific IgG using IgG tracer technology has been shown to be a sensitive and specific assay for the routine diagnostic testing of extrinsic allergic alveolitis (14).
A woman with alveolitis due to her Budgerigar developed recurrent symptoms when exposed to dropping allergens left on her artificial Christmas tree (15).
References
  1. van Toorenenbergen AW, Gerth van Wijk R, van Dooremalen G, Dieges PH. Immunoglobulin E antibodies against budgerigar and canary feathers. Int Arch Allergy Appl Immunol 1985;77(4):433-7
  2. de Maat-Bleeker F, van Dijk AG, Berrens L. Allergy to egg yolk possibly induced by sensitization to bird serum antigens. Annals of Allergy 1985;54(3):245-8.
  3. van Toorenenbergen AW, Huijskes-Heins MI, Gerth van Wijk R. Different pattern of IgE binding to chicken egg yolk between patients with inhalant allergy to birds and food-allergic children. Int Arch Allergy Immunol 1994;104(2):199-203.
  4. Gerth Van Wijk R, Van Toorenenbergen AW, Dieges PH. Nasal allergy to avian antigens. Clin Allergy 1987;17(6):515-21
  5. Bosenecker A, Musken H, Bergmann KC. Sensitization in budgerigar owners. [German] Pneumologie 1998;52(4):209-13
  6. Tauer-Reich I, Fruhmann G, Czuppon AB, Baur X. Allergens causing bird fancier’s asthma. Allergy 1994;49(6):448-53
  7. Martinez-Cordero E, Aquilar Leon DE, Retana VN. IgM antiavian antibodies in sera from patients with pigeon breeder’s disease. J Clin Lab Anal 2000:14(5):201-7
  8. Yoshizawa Y, Miyashita Y, Inoue T, Sumi Y, Miyazaki Y, Sato T, et al. Sequential evaluation of clinical and immunological findings in hypersensitivity pneumonitis: serial subclass distribution of antibodies. Clin immunol Immunopathol 1994;73(3):330-7
  9. Todd A, Coan R, Allen A. Pigeon breeder’s lung; IgG subclasses to pigeon Intestinal mucin and IgA antigens. Clin Exp Immunol 1993;92(3):494-9
  10. Baldwin CI, Todd A, Bourke SJ, Allen A, Calvert JE. IgG subclass responses to pigeon intestinal mucin are related to development of pigeon fancier’s lung. Clin Exp Allergy 1998;28(3):349-57
  11. McSharry C, Macleod K, McGregor S, Speekenbrink AB, Sriram S, Boyd F, et al. Mucosal immunity in extrinsic allergic alveolitis: salivary immunoglobulins and antibody against inhaled avian antigens among pigeon breeders. Clin Exp Allergy 1999;29(7):957-64
  12. Rodrigo MJ, Benavent MI, Cruz MJ, Rosell M, Murio C, Pascual C, et al. Detection of specific antibodies to pigeon serum and bloom antigens by enzyme linked immunosorbent assay in pigeon breeder’s disease. Occup Environ Med 200057(3):159-64
  13. Rodriguez de Castro F, Carrillo T, Castillo R, Blanco C, Diaz F, Cuevas M. Relationship between characteristics of exposure to pigeon antigens. Clinical manifestations and humoral immune response. Chest 1993;103(4):1059-63
  14. Lopata A, Schinkel M, Andersson C, Johansson G, van Hage-Hamsten M. Quantification of IgG antibodies to bird antigens in the diagnosis of extrinsic allergic alveolitis (EAA) using the UniCAP system. (Manuscript in preparation)
  15. Baverstock AM, White RJ. A hazard of Christmas: Bird Fancier's Lung and the Christmas tree. Respir Med 2000;94(2):176