Whole Allergen
e83 Swine epithelium
e83 Swine epithelium Scientific Information
Type:
Whole Allergen
Display Name:
Swine epithelium
Allergen code:
e83
Family:
Suidae
Species:
Sus scrofa
Route of Exposure:
inhalation
Latin Name:
Sus scrofa
Summary
Contact with swine epithelium or derived proteins occurs mainly during swine farming. Animal rehabilitation facilities, veterinary practice, owning pigs as pets, and contact with wild boars also allow contact with S. scrofa epithelium.  Swine farming is frequently associated with adverse respiratory effects including rhinoconjunctivitis, asthma, decreased lung function, however, IgE sensitization to swine epithelium is infrequent. Similarly, hypersensitivity pneumonitis associated with swine farming is seldom associated with swine epithelial antigens. IgE sensitization to swine epithelium has been proposed as a marker of endocarditis on cardiac valve bioprosthesis.
Allergen
Nature
Swine epithelium.
Taxonomy
Species S. scrofa comprises domestic swine (S. scrofa domesticus) and wild boar (S. scrofa) [1].
Taxonomic table 
Domain
Eukaryota
Kingdom
Metazoa (Animalia)
Phylum
Chordata
Subphylum
Vertebrata
Class
Mammalia
Order
Artiodactyla
Family
Suidae
Genus
Sus
Species
S. scrofa
Tissue
Swine epithelium (skin).
Epidemiology
Risk factors 
Unlike cat, dog, or horse, S. scrofa is infrequently kept as a family pet [2]. Therefore, contact with swine epithelium occurs mainly as a result of occupational exposure: swine farming, veterinarians, other agricultural workers [3]. The increase in unconventional pet keeping may also result in increased contact with S. scrofa epithelium [4]. Conversely, despite wild boar hunting being practiced worldwide and boar farming receiving increasing interest [1], no reported allergy to boar epithelium has been reported yet.  Porcine bioprosthesis for heart valve replacement has been associated with IgE sensitization to swine epithelium and in rare cases to noninfectious endocarditis attributed to swine allergy [5, 6]. 
Route of Exposure
Main
The main route of exposure to swine epithelium is through inhalation of allergenic molecules [3]. 
Secondary
Direct skin contact with swine epithelium and surgical implantation of porcine bioprosthetic materials are alternative routes of exposure [3, 5]. 
Clinical Relevance
Despite swine farming being associated with adverse respiratory conditions, IgE sensitization to swine epithelium is an infrequent finding, the main pathophysiological culprit being IgE-independent mechanisms [3].
Allergic reactions to porcine bioprostheses for valvular replacement have been associated to endocarditis [5, 6].
Molecular Aspects
Allergenic molecules
Porcine serumalbumin Sus s 1 is the only swine allergen included in the IUIS/WHO database as of June 2022 [7]. Allergens of the serumalbumin family including Sus s 1 are found in meat and on epithelia,  display cross-reactivity and cannot be used as allergen markers for genuine sensitization [8].
Name
Biochemical activity
Molecular weight (kDa)
Glycosylation
Major/minor allergen
Marker allergen
IUIS
Reference
Sus s 1
swine serumalbumine
60
Minor
No
Yes
[7, 8]
Cross-reactivity
There is partial cross-reactivity between swine hair and dander and hair, dander and serum from other domestic animals and pets [9]. Swine epithelium is not involved in the so-called pork-cat syndrome, which is due to allergen cross-reactivity between cat dander and pork meat [8].
Compiled By
Author: Joana Vitte
Reviewer: Dr. Christian Fischer
Last reviewed: June 2022.
References
  1. Sales, J. and R. Kotrba, Meat from wild boar (Sus scrofa L.): a review. Meat Sci, 2013. 94(2): p. 187-201.
  2. Curin, M. and C. Hilger, Allergy to pets and new allergies to uncommon pets. Allergol Select, 2017. 1(2): p. 214-221.
  3. Sigsgaard, T., et al., Respiratory diseases and allergy in farmers working with livestock: a EAACI position paper. Clin Transl Allergy, 2020. 10: p. 29.
  4. Diaz-Perales, A., et al., Allergy to uncommon pets: new allergies but the same allergens. Front Immunol, 2013. 4: p. 492.
  5. Loyens, M., et al., Link between endocarditis on porcine bioprosthetic valves and allergy to pork. International Journal of Cardiology, 2013. 167(2): p. 600-602.
  6. Fournier, P., et al., A deadly aversion to pork. Lancet, 2011. 377(9776): p. 1542-1542.
  7. IUIS/WHO. IUIS/WHO Chordata.  June 12 2022]; Available from: http://allergen.org/search.php?allergenname=&allergensource=cypress&TaxSource=Animalia+Chordata&TaxOrder=&foodallerg=all&bioname=.
  8. Matricardi, P.M., et al., EAACI Molecular Allergology User's Guide. Pediatr Allergy Immunol, 2016. 27 Suppl 23: p. 1-250.
  9. Prahl, P., B. Weeke, and H. Lowenstein, Quantitative immunoelectrophoretic analysis of extract from cow hair and dander. Characterization of the antigens and identification of the allergens. Allergy, 1978. 33(5): p. 241-53.