Table of Contents
Component
f448 Jug r 2
f448 Jug r 2 Scientific Information
Type:
Component
Name; WHO/IUIS:
Jug r 2
Biological function:
Seed storage protein
Allergen code:
f448
Molecular Weight:
5-7 kDa and 48/145 kDa (monomer/trimer)
Route of Exposure:
Ingestion
Source Material:
English walnut (Juglans regia)
Other Names :
7S globulin (vicilin)
Allergen
Summary
Jug r 2 is a 7S globulin (vicilin) seed storage protein from English walnut (Juglans regia). It is a member of the cupin superfamily and notable for its heat and proteolytic stability. Jug r 2 acts as a primary sensitizing walnut allergen and a marker of genuine walnut allergy rather than pollen-related cross-reactivity. Sensitization to Jug r 2 is associated with systemic, often severe reactions, including anaphylaxis, and is especially relevant in patients with early-onset walnut allergy.
Epidemiology
Walnut is among the most clinically relevant causes of tree nut allergies worldwide. According to data from the EuroPrevall project, 3% of the adult European population is predicted to be sensitized to walnut, ranging from 0.1% in Iceland to 6% and 8% in Switzerland and Spain, respectively (Burney 2014). Sensitization to walnut among children across Europe ranges from 1.4% in Iceland to 9.5% in Switzerland (Lyons 2020).
In a systematic review and meta-analysis (Spolidoro 2024), walnut allergy and sensitization were assessed in 12 studies, of which 11 contributed data to the pooled sensitization analyses. Across these studies, the prevalence of walnut specific IgE sensitization was 4.1% and 2.7% for skin prick testing. In contrast, the pooled estimate for food challenge–confirmed walnut allergy was 0.02%, based on one available study.
According to telephone interviews with 4,855 randomly selected households in the United States, 62% of participants (51/82) who had information about their specific tree nut allergies, reported having walnut allergy (Sicherer 2003).
Data from the European Anaphylaxis Registry, collected between 2007 and 2015, showed that among children and adolescents with anaphylaxis to tree nuts, walnuts were one of the most significant triggers, accounting for 16% (42/256) of the cases (Grabenhenrich 2016).
Sensitization to Jug r 2 and other walnut storage proteins is associated with early-onset walnut allergy in childhood and correlates with severe reactions (Ballmer-Weber 2019, Lyons 2021).
In Ballmer-Weber et al. (2019), vicilin sensitization (Jug r 2 and Jug r 6) was consistently frequent across all European regions studied (Switzerland, Germany, Spain). Unlike other allergens with regionally skewed patterns—such as Jug r 3 (nsLTP), dominant in Spain, or Jug r 5 (PR-10), dominant in birch-endemic central Europe—the vicilin allergens Jug r 2/Jug r 6 showed no strong regional variation. This indicates that Jug r 2 and Jug r 6 act as primary sensitizers across regions with different climates and pollen exposure.
Environmental Characteristics
Source and Tissue
Jug r 2 is a seed storage protein expressed in the cotyledons of the walnut kernel, where it contributes to nutrient reserves during germination. It is a 7S globulin (vicilin) forming trimeric complexes (~145 kDa). Walnut belongs to the Juglandaceae family, and Jug r 2 shares conserved cupin motifs with homologous proteins in other nuts (Barre 2008, Scala 2018).
Risk Factors
Risk factors for Jug r 2 sensitization include early introduction of walnut, an atopic background or concurrent tree nut allergy, and IgE cross-reactivity via structurally related vicilins from closely related Juglandaceae species - such as black walnut (Jug n 2) and pecan (Car i 2) - which share high sequence identity with Jug r 2 (Costa 2014).
Clinical Relevance
Jug r 2 is a clinically important allergen linked to severe systemic reactions, including anaphylaxis (Teuber 1999, Pastorello 2004, Dubiela 2018, Hausmann 2023). In a prospective multicentre study of 115 European patients assessing walnut allergy severity and component resolved sensitization across age groups and regions, vicilin fractions containing Jug r 2 and Jug r 6 fragments correlated significantly with severe symptoms in children (Ballmer-Weber 2019). The same study also reported that sensitization to those vicilin fractions is closely associated with early-onset of walnut allergy.
The prevalence of Jug r 2 sensitization varies dependent on patient population. Early work by Teuber (1999) identified Jug r 2 as a major allergen, with IgE recognition in 60% (9/15) of patients from USA with severe systemic walnut allergy. In contrast, among an Italian cohort predominantly characterised by LTP-driven allergy, vicilin sensitization was detected in 22% (10/46) of patients (Pastorello 2004). Similarly, in a small group UK/Spain cohort with confirmed history of walnut allergy, 27% (3/11) showed IgE-reactivity to 7S globulin fragments (Downs 2014).
Small amounts of walnut can trigger severe reactions in patients’ monosensitized to Jug r 2 as illustrated by a recent case report (Hausmann 2023). The patient experienced anaphylaxis within 30 mins after ingestion of a salad from which a walnut topping had been removed. Walnut extract IgE was 4.98 kUA/l and component testing showed Jug r 2–specific IgE of 8.58 kUA/L. Specific IgE was <0.10 kUA/L for Jug r 1, Jug r 3, Jug r 6, and 0.29 kUA/L for Jug r 4.
Diagnostic Relevance
Component-resolved diagnosis using Jug r 2 can improve discrimination between primary walnut allergy and birch pollen–related cross-reactivity (Ballmer-Weber 2019). Specific IgE to Jug r 2 also appears to be a useful marker for identifying patients at increased risk of severe reactions (Pastorello 2004, Costa 2014, Ballmer-Weber 2019). In addition, Jug r 2 sensitization is associated with early-onset walnut allergy, with significantly higher sensitization rates in patients whose symptoms began in childhood (<14 years) (Ballmer-Weber 2019), providing valuable clinical information in the context of paediatric and early-onset disease.
Prevention and Therapy
There is currently no regulatory approved walnut oral immunotherapy available. Management involves avoidance of walnut and often cross-reactive nuts (especially pecan), education and epinephrine auto-injector prescription when indicated.
In a clinical study including 73 walnut allergic patients it was demonstrated that walnut oral immunotherapy can induce desensitisation to walnut as well as cross-desensitisation to pecan and, in some cases, hazelnut in patients who have tree nut co-allergies (Elizur 2019).
Molecular Aspects
Jug r 2 is a vicilin-type 7S globulin undergoing complex processing during protein maturation, the C-terminal cupin motif-containing domain forming homotrimer assemblies and the N-terminal part being cleaved into six separate hairpinin fragments (Downs 2014). The C-terminal part shares β-barrel cupin domains with other legume and nut vicilins (Barre 2008). Heat and digestion stability explain its persistence as a potent food allergen (Costa 2014, Geiselhart 2018).
The WHO/IUIS Allergen Nomenclature Sub-Committee records the full-length Jug r 2 allergen as Jug r 2.0102.
It also includes two entries corresponding to segments of the Jug r 2.0102 sequence: the N‑terminal hairpining domain (residues 27–367), and the cupin domain (residues 368–789).
In addition, the database lists Jug r 2.0101, which is annotated with an incomplete sequence, and an entry consisting of Jug r 2.0101 residues 1-173.
Cross-reactivity
Inhibition studies demonstrated that walnut pretreatment completely blocks IgE binding to pecan (Elizur 2020) indicating presence of shared epitopes that could lead to 7S globulin-mediated walnut-pecan clinical cross-reactivity. Walnut oral immunotherapy can induce cross-desensitisation to pecan and hazelnut in patients who have tree nut co-allergies (Elizur 2019). IgE cross-reactivity between Jug r 2 and pecan 7S globulin Car i 2, is expected based on their close to 70% amino acid sequence identity (Scala 2018) and presumed high similarity of molecular structures (Barre 2008).
Jug r 2 shares 44% amino acid identity with Jug r 6 vicilin and 30-70% identity with 7S globulins from other tree nuts, such as pecan (Car i 2), hazelnut (Cor a 11), pistachio (Pis v 3), and cashew (Ana o 1). It also shares identity with seeds, such as sesame (Ses i 3) and lupin (Lup an 1), and legumes, such as peanut (Ara h 1) and soybean (Gly m 5) (Dubiela 2018; Scala 2018).
The cupin domains of Jug r 2 and Cor a 16 share 72% sequence identity (personal communication Jonas Lidholm) indicating a strong likelihood for IgE cross-reactivity.
Exposure                                                                                                          
Exposure occurs through ingestion of raw, roasted, or processed walnut products. Jug r 2 is thermostable and resists degradation during cooking or digestion (Costa 2014), explaining reactions to trace amounts of walnut vicilins in baked or confectionery goods (Su 2004, Kim 2018).
Explained Results
Allergen information
Jug r 2 is a 7S globulin (vicilin) seed storage protein. It is heat- and digestion-stable and functions as a marker of genuine walnut allergy.
Clinical relevance
Jug r 2 sensitization is linked to severe systemic reactions and is closely associated with early-onset walnut allergy. Data suggests that Jug r 2 acts as a primary sensitizing walnut allergen across regions with climates and pollen exposure.
Cross-reactivity
Jug r 2 has close to 70% amino acid sequence identity with pecan vicilin Car i 2 that is expected to lead to clinical cross-reactivity. Jug r 2 also show around 40% identity with vicilins from other tree nuts, seeds and legumes.
Compiled by Dr. Michael Spangfort
Reviewed by Dr. Jonas Lidholm, December 2025
References
Ballmer-Weber BK, Lidholm J, Lange L, Pascal M, Lang C, Gernert S, Lozano-Blasco J, Gräni N, Guillod C, Wangorsch A, Hanschmann KM, Pontoppidan B, Tjäder L, Bartra J, Vieths S. Allergen Recognition Patterns in Walnut Allergy Are Age Dependent and Correlate with the Severity of Allergic Reactions. J Allergy Clin Immunol Pract. 2019;7:1560-1567.
Barre A, Sordet C, Culerrier R, Rancé F, Didier A, Rougé P. Vicilin allergens of peanut and tree nuts (walnut, hazelnut and cashew nut) share structurally related IgE-binding epitopes. Mol Immunol. 2008;45:1231-40.
Burney PG, Potts J, Kummeling I, Mills EN, Clausen M, Dubakiene R, Barreales L, Fernandez-Perez C, Fernandez-Rivas M, Le TM, Knulst AC, Kowalski ML, Lidholm J, Ballmer-Weber BK, Braun-Fahlander C, Mustakov T, Kralimarkova T, Popov T, Sakellariou A, Papadopoulos NG, Versteeg SA, Zuidmeer L, Akkerdaas JH, Hoffmann-Sommergruber K, van Ree R. The prevalence and distribution of food sensitization in European adults. Allergy. 2014;69:365-71.
Costa J, Carrapatoso I, Oliveira MBPP, Mafra I. Walnut allergens: molecular characterization, detection and clinical relevance. Clinical & Experimental Allergy. 2014;44, 319–341.
Downs ML, Semic-Jusufagic A, Simpson A, Bartra J, Fernandez-Rivas M, Rigby NM, Taylor SL, Baumert JL, Mills EN. Characterization of low molecular weight allergens from English walnut (Juglans regia). J Agric Food Chem. 2014; 3;62:11767-75.
Dubiela P, Kabasser S, Smargiasso N, Geiselhart S, Bublin M, Hafner C, Mazzucchelli G, Hoffmann-Sommergruber K. Jug r 6 is the allergenic vicilin present in walnut responsible for IgE cross-reactivities to other tree nuts and seeds. Sci Rep. 2018;8:11366.
Elizur A, Appel MY, Nachshon L, Levy MB, Epstein-Rigbi N, Pontoppidan B, Lidholm J, Goldberg MR. Walnut oral immunotherapy for desensitisation of walnut and additional tree nut allergies (Nut CRACKER): a single-centre, prospective cohort study. Lancet Child Adolesc Health. 2019;3312-321.
Elizur A, Appel MY, Nachshon L, Levy MB, Epstein-Rigbi N, Pontoppidan B, Lidholm J, Goldberg MR. Clinical and Molecular Characterization of Walnut and Pecan Allergy (NUT CRACKER Study). J Allergy Clin Immunol Pract. 2020;8:157-165.
Geiselhart S, Hoffmann-Sommergruber K, Bublin M. Tree nut allergens. Mol Immunol. 2018;100:71-81.
Grabenhenrich LB, Dölle S, Moneret-Vautrin A, Köhli A, Lange L, Spindler T, Ruëff F, Nemat K, Maris I, Roumpedaki E, Scherer K, Ott H, Reese T, Mustakov T, Lang R, Fernandez-Rivas M, Kowalski ML, Bilò MB, Hourihane JO, Papadopoulos NG, Beyer K, Muraro A, Worm M. Anaphylaxis in children and adolescents: The European Anaphylaxis Registry. J Allergy Clin Immunol. 2016;137:1128-1137.
Hausmann O, Lidholm J, Ballmer-Weber B. EAACI Congress Hamburg 2023. Poster No. 100382.
Kim JH, Hong JY, Moon JC, Kwon K, Jang CS. Development of molecular markers for detecting almond, peanut, pine nut, and walnut in commercial food using quantitative real-time PCR. Appl Biol Chem. 2018;61:345–354.
Lyons SA, Clausen M, Knulst AC, Ballmer-Weber BK, Fernandez-Rivas M, Barreales L, Bieli C, Dubakiene R, Fernandez-Perez C, Jedrzejczak-Czechowicz M, Kowalski ML, Kralimarkova T, Kummeling I, Mustakov TB, Papadopoulos NG, Popov TA, Xepapadaki P, Welsing PMJ, Potts J, Mills ENC, van Ree R, Burney PGJ, Le TM. Prevalence of Food Sensitization and Food Allergy in Children Across Europe. J Allergy Clin Immunol Pract. 2020;8:2736-2746.
Lyons SA, Datema MR, Le TM, Asero R, Barreales L, Belohlavkova S, de Blay F, Clausen M, Dubakiene R, Fernández-Perez C, Fritsche P, Gislason D, Hoffmann-Sommergruber K, Jedrzejczak-Czechowicz M, Jongejan L, Kowalski ML, Kralimarkova TZ, Lidholm J, Papadopoulos NG, Pontoppidan B, Popov TA, Prado ND, Purohit A, Reig I, Seneviratne SL, Sinaniotis A, Vassilopoulou E, Versteeg SA, Vieths S, Zwinderman AH, Welsing PMJ, Mills ENC, Ballmer-Weber BK, Knulst AC, Fernández-Rivas M, Van Ree R. Walnut Allergy Across Europe: Distribution of Allergen Sensitization Patterns and Prediction of Severity. J Allergy Clin Immunol Pract. 2021;9:225-235.
Pastorello EA, Farioli L, Pravettoni V, Robino AM, Scibilia J, Fortunato D, Conti A, Borgonovo L, Bengtsson A, Ortolani C. Lipid transfer protein and vicilin are important walnut allergens in patients not allergic to pollen. J Allergy Clin Immunol. 2004;114:908-914.
Scala E, Villalta D, Meneguzzi G, Giani M, Asero R. Storage molecules from tree nuts, seeds and legumes: relationships and amino acid identity among homologue molecules. Eur Ann Allergy Clin Immunol. 2018;50:148-155.
Sicherer SH, Muñoz-Furlong A, Sampson HA. Prevalence of peanut and tree nut allergy in the United States determined by means of a random digit dial telephone survey: a 5-year follow-up study. J Allergy Clin Immunol. 2003;112:1203-1207.
Spolidoro GCI, Lisik D, Nyassi S, Ioannidou A, Ali MM, Amera YT, Rovner G, Khaleva E, Venter C, van Ree R, Worm M, Vlieg-Boerstra B, Sheikh A, Muraro A, Roberts G, Nwaru BI. Prevalence of tree nut allergy in Europe: A systematic review and meta-analysis. Allergy. 2024;79:302-323.
Su M, Venkatachalam M, Teuber, SS, Roux KH, Sathe SK. Impact of c-irradiation and thermal processing on the antigenicity of almond, cashew nut and walnut proteins. J Sci Food Agric 2004;84:1119–1125.
Teuber SS, Jarvis KC, Dandekar AM, Peterson WR, Ansari AA. Identification and cloning of a complementary DNA encoding a vicilin-like proprotein, jug r 2, from english walnut kernel (Juglans regia), a major food allergen. J Allergy Clin Immunol. 1999;104:1311-1320.