Whole Allergen
f274 Marjoram
f274 Marjoram Scientific Information
Type:
Whole Allergen
Display Name:
Marjoram
Allergen code:
f274
Family:
Lamiaceae
Latin Name:
Origanum majorana
Other Names:
Marjoram, Sweet Marjoram
Environmental Characteristics
The sub-bushy plant Marjoram originated in the southern Mediterranean region and spread to southern Europe very early. For the Greeks, it was sacred, made into wreaths for weddings and funerals.
Marjoram is a member of the Lamiaceae or Mint family, which includes many familiar culinary herbs: Mint, Basil, Oregano and Sage. Sweet Marjoram is more compact and typically grows in a “mound”, with branches spilling over and taking root where they touch the ground, whereas many kinds of Oregano are upright. Marjoram flowers tend to be white, and the leaves smaller (only up to 2.5 cm) and grayish-green and softer-textured. They have a distinctive fragrance and taste (but less strong than those of Oregano), which give them broad culinary uses. Marjoram is rather delicate and is classified as a summer annual in most regions, in contrast to Oreganos in general, which are held to be perennials.
Marjoram is most often used in recipes of French or English origin, whereas Oregano's more robust flavour is habitually called for in Italian, Greek, North African and Mexican cuisines. Marjoram is far from unknown in Mediterranean cooking, however. It is not strongly associated with particular dishes; rather, it is a familiar ingredient of many soups and stews, and often flavours poultry and seafood. It frequently features in blends of herbs, and can be made into a tea.
The essential oil comprises between 0.7% and 3.5%. The main aroma component is a bicyclic monoterpene alcohol, cis-sabinene hydrate. Oil of Marjoram is used in perfumes. Medicinal uses include pain relief and the treatment of digestive complaints.
Clinical Relevance
IgE-mediated reactions
Marjoram may uncommonly induce symptoms of food allergy in sensitised individuals. As little has been reported in the literature, adverse reactions to the closely related family member Oregano will be of help.
A 45-year-old man experienced 3 reactions to food: as a result of Oregano on a single occasion, and twice to Thyme. He had pruritis and swelling of the lips and tongue, dysphagia, dysphonia, and progressive upper respiratory difficulty, as well as intense facial and palpebral oedema. On 2 occasions he also had hypotension, vomiting, and nausea. Onset was within minutes after the ingestion of pizza containing the herb in the first instance, meat seasoned with Thyme in the second, and snails with Thyme in the third (1).
A 45-year-old female patient, with facial eczema that appeared 20 minutes after ingestion of Oregano and was exacerbated by sun exposure, was reported. A patch test was positive. Three other patients were reported to be positive to Oregano on a patch test, but their symptoms were not described (2).
Perioral dermatitis following ingestion of Marjoram has been reported (3).
Molecular Aspects
No allergens from this plant have yet been characterised.
Cross Reactivity
An extensive cross-reactivity among the different individual species of the genus could be expected, and in particular between Oregano and Marjoram (4).
Compiled By
Last reviewed : June 2022
References
  1. Benito M, Jorro G, Morales C, Pelaez A, Fernandez A. Labiatae allergy: systemic reactions due to ingestion of oregano and thyme. Ann Allergy 1996;76(5):416-8
  2. Futrell JM, Rietschel RL. Spice allergy evaluated by results of patch tests. Cutis 1993;52(5):288-290
  3. Farkas J. Perioral dermatitis from marjoram, bay leaf and cinnamon. Contact Dermatitis 1981;7(2):121
  4. Yman L. Botanical relations and immunological cross-reactions in pollen allergy. 2nd ed. Pharmacia Diagnostics AB. Uppsala. Sweden. 1982: ISBN 91-970475-09