Whole Allergen
f31 Carrot
f31 Carrot Scientific Information
Type:
Whole Allergen
Display Name:
Carrot
Allergen code:
f31
Family:
Apiaceae
Species:
carota
Route of Exposure:
Ingestion
Latin Name:
Daucus carota
Other Names:
Bird's nest, Bishop's lace, Queen Anne's lace, Wild carrot
WHO/ICD-11 code:
XM9S46
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Summary
Carrot (Daucus carota) is one of the most important, widely cultivated root vegetables, belonging to Apiaceae family with a rank in the top 10 vegetables around the world. The most widely cultivated species of carrot include D. carota L. subsp. sativus with Central Asia to be considered as the origin. They are classified as eastern and western carrots based on the pigmentation of the roots with colors ranging from orange, yellow, purple, red and white. The fleshy edible taproots of carrots may be allergic to some individuals. Carrot sensitization was found wide across Europe and even in the USA, Mexico, and certain countries of Asia. Carrot is one of the prominent triggering vegetables for pollen-food-allergy-syndrome associated with birch and mugwort pollens. Ingestion can trigger oral allergy syndrome or pollen food allergy syndrome with symptoms ranging from swelling of lips, tongue and throat, nausea, dysphonia, itching of the nose and ear to even systemic reactions like anaphylaxis in some. Dau c 1 reported as a major allergen, out of the 4 allergens, which is homologous to Bet v 1, the birch pollen allergen. Carrot allergens are highly cross-reactive with especially birch tree pollens and celery. The allergy might be prevented in carrot sensitized people by avoiding any intentional or un-intentional consumption of carrot.
Allergen
Nature
Carrot (Daucus carota) is one of the most important root vegetables in the Apiaceae family and is widely cultivated around the world with a rank in the top 10 vegetables (1, 2). It is a biennial erect herbaceous plant with 20-50 cm tall during the mature vegetative stage and 120-150 cm tall when flowering (3). The part of the plant that is mostly consumed is the roots due to its richness in carotenoids, anthocyanins, dietary fiber, vitamins and other nutrients (2). Carrot plants flowers in the second year of growth. Carrot flowers are flattened umbrella-shaped umbel and usually white in color. The leaves of the carrot plants are glabrous, green compound leaves growing in a rosette (2).
Habitat
Carrot is found in both wild and cultivated types. Wild carrot is a weed and can spread very fast. The cultivated carrot is mostly used as a vegetable. Carrot grows in natural grasslands or cultivated lands (3).
Taxonomy
The taxonomic classification of the genus Daucus is not clear. Earlier studies subdivided the genus into 5 sections of 20 species with Daucus L. as one having 12 species. D. carota subsp. carota is the best-known wild species among them. The most widely cultivated carrot includes D. carota L. subsp. sativus domesticated from wild species (4). Spooner et. al., performed a multivariate analysis among D. carota species and proposed to classify all of the known subspecies under two main subspecies as D. carota subsp. carota and subsp. gummifer (4, 5). The cultivated carrots are classified into eastern carrots and western carrots based on the pigmentation of roots. Eastern carrot roots are branched, purple or yellow in color due to the presence of anthocyanin with slightly dissected leaves. Western carrot roots are unbranched, orange-red or white in color due to the presence of carotene with highly dissected leaves (2).
Epidemiology
Worldwide distribution
Most of the carrot allergic persons are reported to be sensitive to birch or mugwort pollens (6). A survey regarding triggers and symptoms of food allergy was conducted in Germany on 73 patients with a history of birch pollen associated food allergy. The results reported that the carrot was responsible for eliciting allergic reactions in 52% of patients (7). According to a EuroPrevall analysis (the study about the basis and prevalence of food allergy across Europe), 3.6% prevalence of sensitization to carrot was observed in a study on 4522 adults (aged 20-39 years) from 13 countries across Europe (8). Apple, banana and carrot were triggering foods for 5% of pollen food allergy syndrome (PFAS) due to birch and ragweed pollens in 122 children (0-18 year) in the USA (9), whereas, apple, peach and carrot were triggering foods for 26.7% of PFAS due to birch, grass and ragweed pollens in Croatia including 120 patients (10). A total of 2633 Mexican subjects were tested for food allergy through immunoglobulin E (IgE) positivity and carrot was found as food with high positivity in 37.5% of subjects although less than 50 samples tested (11).
A prevalence study of self-reported food allergy (FA), food sensitization (FS) and probable FA (individuals having symptoms plus IgE sensitization) to 24 Europrevall priority foods in adults (12) and in school-age children across Europe (13) was conducted to analyze geographical variation. Probable FA to carrot was markedly common among adults in Zurich, Switzerland (1.0 %) than in other countries of Europe (12). Carrot constituted 1.1% of 16,935 children (7-10 years old) for self-reported FA. Carrot along with hazelnut, apple and celery were important sources of FA in children in Central and Northern Europe. Carrot was among the most common sensitizing food in children causing probable FA in birch-endemic countries, Switzerland, Netherlands, Poland and Lithuania, maybe due to cross-reactivity with birch PR-10 protein (13). The prevalence of FS for carrot in the total study population (2196 patients) with available food serology was found to be 12.4% in Zurich, 8.3% in Madrid, 4.8% in Athens, 7.2% in Utrecht, 4.5% in Vilnius, 5.0% in Lodz and 2.1% in Reykjavik. Carrot was the cause of self-reported FA and matching FS in 30.3% of the total of 1989 subjects with available food serology (13).  
A similar study was conducted in South India for the 24 Europrevall priority foods in 11,791 adults (20-54 years), however, the South-Indian population was not found to be allergic to carrot as compared to high sensitization in Europe (14). According to a study conducted in China, India and Russia for 24 Europrevall priority foods in 35,549 school children (6-11 years), 1.5% subjects in Hongkong, 0% in Guangzhou, 2.0% in Shaoguan, 2.5% in Tomsk and 6.7% in India depicted positive serum specific IgE to carrot. However, only 0.15% of subjects in Russia reported symptoms with positive IgE, while others countries were not found to have a probable food allergy to carrot (15).
Risk factors
Carrot is considered to be one of the prominent triggering vegetables for pollen-food-allergy-syndrome (PFAS) associated with birch and mugwort pollens (10).
Environmental Characteristics
Living environment
Carrot adapts and grows in a cool climate and can be sown in spring in temperate zones or in winter in subtropical zones (2). It grows well at altitudes of 700 m in tropical regions with an optimum temperature of 16-24ºC and grows best in well-drained fertile soil with sandy texture and pH of 6.0 to 6.5 (3).
Worldwide distribution
Central Asia is considered to be one of the origins of cultivated carrot with Afghanistan to be thought as the first center of diversity and Turkey as the second center (2). Its cultivation has spread throughout the world and is now native to temperate regions of Europe, Southwestern Asia, and North Africa. It is also grown in North America, Japan, New Zealand and Australia (3). 
Route of Exposure
Main
Ingestion of carrot can trigger food related allergic response as Oral Allergy Syndrome (OAS) or Pollen food allergy syndrome (PFAS) (16-19).
Clinical Relevance
Food Allergy
Carrot is normally associated with food allergy and OAS or PFAS (Sánchez-Guerrero et al, 2020). Allergic symptoms in OAS mostly include the oropharyngeal regions such as swelling of lips, tongue, and throat, nausea, dysphonia, and itching of the nose and ear (17, 20). About 50% of carrot allergic patients develop systemic reactions (6). In a study on 55 patients with airborne allergy, food hypersensitivity (mainly OAS) was reported in 15 patients especially with apples, carrots, cherry, peach, hazelnut, walnut, celery and nectarines (21). Systemic allergic reactions are reported after the ingestion of carrot in pollen allergic patients due to cross-reactivity (22). PFAS was found to be significantly correlated with root vegetables including carrot in a study conducted on 54 seasonal allergic rhinitis patients (23).
Allergic rhinitis
A study on 2305 representative samples of the Korean population found that more carrot consumption (OR - 2.01; CI, 1.01–1.43; p - 0.10) was associated with increased risk of allergic rhinitis (24). A study enrolled 54 patients with seasonal allergic rhinitis and subjected to skin prick tests and microarray analysis to determine the incidence of PFAS and food tolerance. Carrot showed a positive skin prick test in 29 patients (23).
Asthma
Some patients presented asthma due to handling or ingesting raw carrots which were due to major carrot allergen Dau c 1 according to a small study. 2 patients were found to be sensitized to carrots directly without pollens involved while 1 patient has pollen-related carrot allergy (25).
Atopic Dermatitis
Allergic contact dermatitis by Apiaceae root vegetables is rare, however; few cases of dermatitis have been reported due to carrots (26-28).
Other diseases
Anaphylactic shock has been reported in some case studies in carrot allergic patients (17, 29, 30).
Other topics
As per a study, carrot was the food trigger for PFAS in 15.5% of adults with eosinophilic esophagitis (31).
Prevention and Therapy
Prevention strategies
Avoidance
The major management of carrot allergy in pollen sensitized individuals is to avoid eating the food which poses a positive challenge (22). 
Molecular Aspects
Allergenic molecules
A total of 3 allergens have been formally named from carrot by WHO/IUIS Allergen Nomenclature Sub-committee (32), however, Dau c cyclophilin has also been reported in the literature (33, 34).
Allergen
Biochemical Name
Molecular Weight
Allergenicity
Dau c 1
Pathogenesis-related protein, PR-10, Bet v 1 family member
16 kDa
85% of 26 carrot allergics were sensitized to Dau c 1 (35). 98% of 40 carrot allergic patients were allergic to isoallergen Dau c 1.0104 variant while 65% to Dau c 1.0201 (36, 37).
Dau c 4
Profilin
14 kDa
A relatively smaller number of patients (20%) are sensitive to Dau c 4 (35).
Dau c 5
Isoflavone reductase-like protein (IFR)
33 kDa
 IFR-1 and IFR-2 were found in 6% and 20% of 49 carrot allergics respectively (34).
Dau c
Cyclophilin protein
20 kDa
-
The major carrot allergen is Dau c 1, homologous to Bet v 1, the birch pollen allergen belonging to the class of pathogenesis-related class 10 (PR-10) (6). Profilin was considered as a minor allergen in carrots (38). Carrot cyclophilin (Dau c Cyc) showed IgE antibody binding in carrot allergic patients in Japan. However, the study on 49 carrot allergic patients conducted to evaluate the carrot allergens found sensitization to Dau c Cyc in only one carrot allergic patient (34).  Lipid like transfer protein (LTP) was not found to be a concern for carrot allergic individuals (6). Dau c 1 does not change its structure until higher temperatures are reached (39). As per the study conducted by Jacob, et al., the heat sensitivity of Dau c 1 differs with specific isoallergen and pH. The natural Dau c 1 in carrots is heat stable as cooked carrot extract could generate mediator release. Different isoallergens (nDau c 1, Dau c 1.0301, Dau c 1.0201, Dau c 1.0105 and Dau c 1.0104) were found heat resistant and stable at pH 3 (stomach conditions). Further, Dau c 1.0301 at pH 3 and Dau c 1.0201 at pH 7 unfolds but were able to trigger the mediator response (36).
Dau c 1 can induce allergic responses independent from Bet v1. This was proved as Dau c 1 was able to trigger Bet v 1 independent T-cell response in 31 birch pollen-allergic patients showing allergic symptoms after ingestion of raw carrots (40).
Cross-reactiviy
A number of studies have reported cross-reactivity between pollens (especially Birch and mugwort) and carrot (17, 41, 42) leading to oral allergy syndrome.
Cross-reactivity is also found between the species of Apiaceae family too. The major allergen in carrot (Dau c 1) is homologous to Bet v 1 – the major birch pollen allergen. A study has shown that 70% of patients allergic to birch pollen may also be allergic to apples, nuts, celery, soybeans in addition to carrots (43).
Carrot-celery-mugwort spice syndrome is quite known in Europe. Carrot, celery and spices were found to be highly cross-reactive along with sensitivity to birch pollens (44). 
Compiled By
Author: Turacoz Healthcare Solutions
Reviewer: Dr. Fabio Iachetti
Last reviewed: November  2020
References
  1. Ahmad T, Cawood M, Iqbal Q, Ariño A, Batool A, Tariq RMS, et al. Phytochemicals in Daucus carota and Their Health Benefits-Review Article. Foods. 2019;8(9).
  2. Que F, Hou X-L, Wang G-L, Xu Z-S, Tan G-F, Li T, et al. Advances in research on the carrot, an important root vegetable in the Apiaceae family. Horticulture Research. 2019;6(1):69.
  3. Daucus carota [original text by Andrew Praciak [Internet]. CAB International. 2019 [cited October 22, 2020]. Available from: https://www.cabi.org/isc/datasheet/18018.
  4. Arbizu C, Ruess H, Senalik D, Simon PW, Spooner DM. Phylogenomics of the carrot genus (Daucus, Apiaceae). Am J Bot. 2014;101(10):1666-85.
  5. Spooner DM, Bouzbida Bc, El Koudrim M, Ghrabi-Gammar Z, Neffati M, Ouabbou H, et al. Reassessment of Practical Subspecies Identifications of the USDA Daucus carota L. Germplasm Collection: Morphological Data. Crop Sci. 2014;54(2):706-18.
  6. Ballmer-Weber B, Hoffmann-Sommergruber K. Molecular Diagnostics of Allergy to Fruits and Vegetables. In: Kleine-Tebbe J, T. J, editors. Molecular Allergy Diagnostics. Switzerland: Springer International Publishing; 2017. p. 275-6.
  7. Beyer S, Franke A, Simon JC, Treudler R. Measurement of health-related quality of life in adult patients with birch pollen-associated food allergy. J Dtsch Dermatol Ges. 2016;14(4):397-404.
  8. Burney P, Summers C, Chinn S, Hooper R, van Ree R, Lidholm J. Prevalence and distribution of sensitization to foods in the European Community Respiratory Health Survey: a EuroPrevall analysis. Allergy. 2010;65(9):1182-8.
  9. Ma S, Sicherer SH, Nowak-Wegrzyn A. A survey on the management of pollen-food allergy syndrome in allergy practices. J Allergy Clin Immunol. 2003;112(4):784-8.
  10. Mastrorilli C, Cardinale F, Giannetti A, Caffarelli C. Pollen-Food Allergy Syndrome: A not so Rare Disease in Childhood. Medicina (Kaunas). 2019;55(10).
  11. Ruiz Segura LT, Figueroa Pérez E, Nowak-Wegrzyn A, Siepmann T, Larenas-Linnemann D. Food allergen sensitization patterns in a large allergic population in Mexico. Allergol Immunopathol (Madr). 2020.
  12. Lyons SA, Burney PGJ, Ballmer-Weber BK, Fernandez-Rivas M, Barreales L, Clausen M, et al. Food Allergy in Adults: Substantial Variation in Prevalence and Causative Foods Across Europe. J Allergy Clin Immunol Pract. 2019;7(6):1920-8.e11.
  13. Lyons SA, Clausen M, Knulst AC, Ballmer-Weber BK, Fernandez-Rivas M, Barreales L, et al. Prevalence of Food Sensitization and Food Allergy in Children Across Europe. J Allergy Clin Immunol Pract. 2020;8(8):2736-46.e9.
  14. Mahesh PA, Wong GW, Ogorodova L, Potts J, Leung TF, Fedorova O, et al. Prevalence of food sensitization and probable food allergy among adults in India: the EuroPrevall INCO study. Allergy. 2016;71(7):1010-9.
  15. Li J, Ogorodova LM, Mahesh PA, Wang MH, Fedorova OS, Leung TF, et al. Comparative Study of Food Allergies in Children from China, India, and Russia: The EuroPrevall-INCO Surveys. J Allergy Clin Immunol Pract. 2020;8(4):1349-58.e16.
  16. Ballmer-Weber BK, Weber JM, Vieths S, Wüthrich B. Predictive value of the sulfidoleukotriene release assay in oral allergy syndrome to celery, hazelnut, and carrot. J Investig Allergol Clin Immunol. 2008;18(2):93-9.
  17. Muluk NB, Cingi C. Oral allergy syndrome. Am J Rhinol Allergy. 2018;32(1):27-30.
  18. Sánchez-Guerrero IM, Nieto A, Meseguer J, Navarro M, López Sáez MP, Magdalena MK, et al. Occupational Rhinoconjunctivitis Induced by Unusual Allergens of Carrot. J Investig Allergol Clin Immunol. 2020;30(3):204-6.
  19. Wangorsch A, Weigand D, Peters S, Mahler V, Fötisch K, Reuter A, et al. Identification of a Dau c PRPlike protein (Dau c 1.03) as a new allergenic isoform in carrots (cultivar Rodelika). Clin Exp Allergy. 2012;42(1):156-66.
  20. Carlson G, Coop C. Pollen food allergy syndrome (PFAS): A review of current available literature. Ann Allergy Asthma Immunol. 2019;123(4):359-65.
  21. Gocki J, Kołodziejczyk J, Przybyszewski M, Bartuzi Z. [Food hypersensitivity in patients with pollen allergy]. Przegl Lek. 2016;73(12):809-12.
  22. Werfel T, Asero R, Ballmer-Weber BK, Beyer K, Enrique E, Knulst AC, et al. Position paper of the EAACI: food allergy due to immunological cross-reactions with common inhalant allergens. Allergy. 2015;70(9):1079-90.
  23. Ludman S, Jafari-Mamaghani M, Ebling R, Fox AT, Lack G, Du Toit G. Pollen food syndrome amongst children with seasonal allergic rhinitis attending allergy clinic. Pediatr Allergy Immunol. 2016;27(2):134-40.
  24. Rhee CS, Wee JH, Ahn JC, Lee WH, Tan KL, Ahn S, et al. Prevalence, risk factors and comorbidities of allergic rhinitis in South Korea: The Fifth Korea National Health and Nutrition Examination Survey. Am J Rhinol Allergy. 2014;28(2):e107-14.
  25. Moreno-Ancillo A, Gil-Adrados AC, Cosmes PM, Domínguez-Noche C, Pineda F. Role of Dau c 1 in three different patterns of carrot-induced asthma. Allergol Immunopathol (Madr). 2006;34(3):116-20.
  26. Kawai M, Tamagawa-Mineoka R, Hagura A, Masuda K, Katoh N. Allergic contact dermatitis due to carrots. J Dermatol. 2014;41(8):753-4.
  27. Paulsen E, Petersen TH, Fretté XC, Andersen KE, Christensen LP. Systemic allergic dermatitis caused by Apiaceae root vegetables. Contact Dermatitis. 2014;70(2):98-103.
  28. White JML, Pink AE. English Sunday lunch dermatitis: Allergic contact dermatitis to parsnip, carrot, fennel (and ivy). Contact Dermatitis. 2020.
  29. Hernández-Moreno KE, Diez LS. [Systemic reaction after performing a food prick-to-prick test. A case report]. Rev Alerg Mex. 2017;64(1):126-9.
  30. Schiappoli M, Senna G, Dama A, Bonadonna P, Crivellaro M, Passalacqua G. Anaphylaxis due to carrot as hidden food allergen. Allergol Immunopathol (Madr). 2002;30(4):243-4.
  31. Letner D, Farris A, Khalili H, Garber J. Pollen-food allergy syndrome is a common allergic comorbidity in adults with eosinophilic esophagitis. Dis Esophagus. 2018;31(2).
  32. Allergen Nomenclature "Daucus carota - All Allergen." [Internet]. WHO/IUIS Allergen Nomenclature Sub-Committee. 2019 [cited October 22, 2020]. Available from: http://www.allergen.org/search.php?allergensource=Daucus+carota.
  33. Fujita C, Moriyama T, Ogawa T. Identification of cyclophilin as an IgE-binding protein from carrots. Int Arch Allergy Immunol. 2001;125(1):44-50.
  34. Ballmer-Weber BK, Skamstrup Hansen K, Sastre J, Andersson K, Bätscher I, Ostling J, et al. Component-resolved in vitro diagnosis of carrot allergy in three different regions of Europe. Allergy. 2012;67(6):758-66.
  35. Ballmer-Weber BK, Wüthrich B, Wangorsch A, Fötisch K, Altmann F, Vieths S. Carrot allergy: double-blinded, placebo-controlled food challenge and identification of allergens. J Allergy Clin Immunol. 2001;108(2):301-7.
  36. Jacob T, Vogel L, Reuter A, Wangorsch A, Kring C, Mahler V, et al. Food Processing Does Not Abolish the Allergenicity of the Carrot Allergen Dau c 1: Influence of pH, Temperature, and the Food Matrix. Molecular Nutrition & Food Research. 2020;64(18):2000334.
  37. Ballmer-Weber BK, Wangorsch A, Bohle B, Kaul S, Kundig T, Fotisch K, et al. Component-resolved in vitro diagnosis in carrot allergy: does the use of recombinant carrot allergens improve the reliability of the diagnostic procedure? Clin Exp Allergy. 2005;35(7):970-8.
  38. Calamelli E, Liotti L, Beghetti I, Piccinno V, Serra L, Bottau P. Component-Resolved Diagnosis in Food Allergies. Medicina (Kaunas). 2019;55(8).
  39. Lyons SA, Dijk AMV, Knulst AC, Alquati E, Le TM, Os-Medendorp HV. Dietary Interventions in Pollen-Related Food Allergy. Nutrients. 2018;10(10).
  40. Zulehner N, Nagl B, Briza P, Roulias A, Ballmer-Weber B, Zlabinger GJ, et al. Characterization of the T-cell response to Dau c 1, the Bet v 1-homolog in carrot. Allergy. 2017;72(2):244-51.
  41. Waserman S, Bégin P, Watson W. IgE-mediated food allergy. Allergy Asthma Clin Immunol. 2018;14(Suppl 2):55.
  42. Worm M, Jappe U, Kleine-Tebbe J, Schäfer C, Reese I, Saloga J, et al. Food allergies resulting from immunological cross-reactivity with inhalant allergens: Guidelines from the German Society for Allergology and Clinical Immunology (DGAKI), the German Dermatology Society (DDG), the Association of German Allergologists (AeDA) and the Society for Pediatric Allergology and Environmental Medicine (GPA). Allergo J Int. 2014;23(1):1-16.
  43. Vieths S, Scheurer S, Ballmer-Weber B. Current understanding of cross-reactivity of food allergens and pollen. Ann N Y Acad Sci. 2002;964:47-68.
  44. Helbling A, Lopez M, Schwartz HJ, Lehrer SB. Reactivity of carrot-specific IgE antibodies with celery, apiaceous spices, and birch pollen. Ann Allergy. 1993;70(6):495-9.