Whole Allergen
f326 Sheep's whey
f326 Sheep's whey Scientific Information
Type:
Whole Allergen
Display Name:
Sheep's whey
Allergen code:
f326
Latin Name:
Ovis spp.
Summary
Crossreactivity between sheep's whey and cow's milk and whey might be expected. 
Exposure
Allergen Exposure
Sheep's cheese, and other dairy products.
Unexpected exposure
Sometimes included in infant formulas.
Potential Cross Reactivity
Crossreactivity between sheep's whey and cow's milk and whey might be expected.
Milk from other related animals.
Review
Sheep’s whey is sometimes included in infant formulas. Crossreactivity with cow’s milk and whey might be expected.