Whole Allergen
f50 Chub mackerel
f50 Chub mackerel Scientific Information
Type:
Whole Allergen
Display Name:
Chub mackerel
Allergen code:
f50
Family:
Scombridae
Latin Name:
Scomber japonicus
Other Names:
Chub mackerel
WHO/ICD-11 code:
XM95M8
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Summary
This species is very similar to the Atlantic mackerel, Scomber scombrus f206.
Route of Exposure
Chub mackerel is eaten raw or cooked.  This species, which is very similar to the Atlantic mackerel Scomber scombrus, is found mostly in the Pacific, but also in the Atlantic, where it is called a Chub mackerel or, in some languages, Spanish mackerel.
Clinical Relevance
IgE antibodies have been measured to mackerel in two groups of patients with atopic dermatitis (1, 2) and in children (3). Mackerel may contain the parasite Anisakis simplex p4 and presumed sensitivity to mackerel may actually be an allergy to this parasite (4). Members of the Scombroidae family easily form histamine during storage, ("scombroid poisoning").
Other Topics
This species, which is very similar to the Atlantic mackerel Scomber scombrus, is found mostly in the Pacific, but also in the Atlantic, where it is called a Chub mackerel or, in some languages, Spanish mackerel.
IgE antibodies have been measured to mackerel in two groups of patients with atopic dermatitis (1, 2) and in children (3). In Japan, many people appear to be allergic to mackerel but in some cases, this may actually be an allergy to the parasite Anisakis simplex (4). Members of the Scombroidae family easily form histamine during storage, ("scombroid poisoning"). Species within groups of fish, like Gadiformes (examples: codfish and hake) and Scombroid fishes (examples: mackerel and tuna) seem to share allergenic components. The overlap of allergen specificity between the groups seems to be moderate or even small. 
Molecular Aspects
Cross Reactivity
Species within groups of fish, like Gadiformes (examples: codfish and hake) and Scombroid fishes (examples: mackerel and tuna) seem to share allergenic components. The overlap of allergen specificity between the groups seems to be moderate or even small.
Compiled By
Last reviewed : June 2022
References
  1. Matsumaru S, Artia M et al. Clinical evaluation of Pharmacia CAP System new allergens for fish, vegetables, fruits and grains. Jap Soc Ped Allergol 1992.
  2. Lindqvist A, Ikezawa Z, Tanaka A, Yman L. Seafood specific IgE in atopic dermatitis. Ann Allergy 1993;70:58.
  3. Yamada M, Torii S. Clinical evaluation of Pharmacia CAP System new food and inhalant allergens. Japanese Soc Allergol 1992.
  4. Kasuya S, Hamano H, Izumi S. Mackerel-induced urticaria and Anisakis. Lancet 1990;335:665.