Table of Contents
Component
f430 Act d 8, PR-10 protein, Actinidia deliciosa fruit
f430 Act d 8, PR-10 protein, Actinidia deliciosa fruit Scientific Information
Type:
Component
Name; WHO/IUIS:
Act d 8, PR-10 protein, Actinidia deliciosa fruit
Biological function:
Pathogenesis-Related protein (PR-10), Bet v 1-related protein
Allergen code:
f430
Molecular Weight:
17 kDa
Source Material:
recombinant, CCD-free protein
Other Names :
Actinidia deliciosa PR-10 allergen
Summary
Act d 8, an allergen from kiwifruit (Actinidia deliciosa), is a member of the Pathogenesis Related (PR)-10 family, cross-reactive with Bet v 1, the major allergen of birch pollen. Act d 8 is a major kiwifruit allergen in geographical areas subject to birch pollinosis. In patients reacting upon kiwifruit ingestion, Act d 8 sensitization is not associated with severity of the reaction but is inversely correlated to kiwifruit monosensitization. Using Act d 8 specific IgE determination increases the sensitivity of kiwifruit diagnosis compared to skin prick tests or specific IgE to kiwifruit whole allergen extract.
Epidemiology
Worldwide distribution
During the 40 years since the description of the first case, allergy to kiwifruit, usually Actinidia (A.) deliciosa has become a major cause of food and fruit allergy worldwide [1, 2]. In Europe, data collected from 2005 through 2009 as part of the EuroPrevall study placed kiwifruit allergy at the 4th place of most prevalent food allergies in adults from Zurich (Switzerland) and Utrecht (the Netherlands) with a prevalence of 1.34% and 0.57% respectively [2]. In children, the EuroPrevall study found similar prevalence figures, with a maximum of 1.06% in Madrid (Spain) [3]. The prevalence of sensitization to A. deliciosa ranged from 1.8% to 9.4% in both adults and children [2-4]. Kiwifruit was also reported as the most frequent culprit food for pollen-food syndrome in Turkey, reported by 39% of patients [5] and the 3rd most frequent in Korea (30%) [6].
Act d 8 sensitization is part of PR-10 cross-reactivity and therefore displays considerable variation depending on the geographic and climate area considered. In Europe, Act d 8 sensitization was demonstrated in 58% of kiwifruit-allergic patients from Central and Western Europe and in 44% of those from Eastern Europe, but only in 11% of those from birch-free Southern Europe [7].
Environmental Characteristics
Source and tissue
Act d 8 is present in the peripheral pulp of mature green kiwifruit [8].
Risk factors
The main risk factor for developing sensitization to Act d 8 is exposure to birch pollen [9].
Detection methods
Similar to other PR-10 proteins, Act d 8 is a low-abundant, relatively unstable protein which may not be sufficiently represented in whole allergen extracts and therefore lead to insufficient detection sensitivity. Using Act d 8 or a representative allergen such as Bet v 1 in addition to the whole allergen extract improves the diagnostic performance [9].
Clinical Relevance
Act d 8 sensitization is associated with pollen-kiwifruit cross-reactivity and therefore usually manifests itself as an oral allergy syndrome [9]. However, Act d 8 sensitization often occurs in polysensitized patients [7].
Disease severity and prediction
Act d 8 sensitization can coexist with sensitization to other kiwifruit allergens, explaining the lack of a statistically significant association with increased or decreased severity of kiwifruit allergy [7, 10].
Cross-reactive molecules
Act d 8, a member of the PR-10 family of proteins, exhibits cross-reactivity with Bet v 1 and Bet v 1-related allergens [8, 11]. Another kiwifruit allergen, Act d 11, cross-reacts with PR-10 proteins despite low sequence identity and its inclusion in the distinct family of Ripening-Related Proteins /Major Latex Proteins [9, 12].
Prevention and Therapy
Experimental trials
Not relevant.
Molecular Aspects
Biochemistry
Act d 8 is a 157-aminoacid protein displaying the typical PR-10 sequence and structure [8, 13]. It is present in low amounts in the pulp of green kiwifruit [8]. Similar to other PR-10 proteins, Act d 8 is thermolabile, degraded by thermal processing, and relatively unstable, explaining its under-representation in allergenic extracts [9]. 
Isoforms, epitopes, antibodies
As of April 26, 2022, only one isoallergen of Act d 8 has been included in the World Health Organization (WHO) and International Union of Immunological Societies (IUIS) Allergen Nomenclature [14].
Cross-reactivity due to structural similarity
Act d 8 displays moderate sequence identity with other allergenic PR-10 proteins, ranging from 50% to 57% for the main pollen homologues (Bet v 1 53%, Cor a 1 55%, Aln g 1 50%) and from 54% to 61% for the main fruit homologues (Pru p 1 54%, Mal d 1 57%, Pru ar 1 61%). [8]. Studies addressing Act d 8 sensitization in the context of other PR-10 allergens show that Act d 8 is one of the most infrequent PR-10 sensitizations, both in birch-pollen areas and in those without birch pollen [11, 15].
Diagnostic Relevance
Cross-Reactivity
Act d 8 sensitization in kiwifruit-allergic patients is mainly associated with Bet v 1-related cross-reactivity in birch pollen-sensitized patients [9]. Of note, Act d 8-related kiwifruit allergy was reported in patients with birch pollen sensitization but lacking birch pollen-induced symptoms [7].
Exposure
The main route of exposure is through consumption of kiwifruit [9]. 
Compiled By
Author: Joana Vitte
Reviewer: Dr.Christian Fischer
Last reviewed: June 2022
References
  1. Fine, A.J., Hypersensitivity reaction to kiwi fruit (Chinese gooseberry, Actinidia chinensis). J Allergy Clin Immunol, 1981. 68(3): p. 235-7.
  2. Lyons, S.A., et al., Food Allergy in Adults: Substantial Variation in Prevalence and Causative Foods Across Europe. J Allergy Clin Immunol Pract, 2019. 7(6): p. 1920-1928 e11.
  3. Lyons, S.A., et al., Prevalence of Food Sensitization and Food Allergy in Children Across Europe. J Allergy Clin Immunol Pract, 2020. 8(8): p. 2736-2746 e9.
  4. Burney, P.G., et al., The prevalence and distribution of food sensitization in European adults. Allergy, 2014. 69(3): p. 365-71.
  5. Ozdemir, S.K. and S. Ozguvarsigmalu, Pollen food allergy syndrome in Turkey: Clinical characteristics and evaluation of its association with skin test reactivity to pollens. Asian Pac J Allergy Immunol, 2018. 36(2): p. 77-81.
  6. Kim, M.A., et al., Pollen-Food Allergy Syndrome in Korean Pollinosis Patients: A Nationwide Survey. Allergy Asthma Immunol Res, 2018. 10(6): p. 648-661.
  7. Le, T.M., et al., Kiwifruit allergy across Europe: clinical manifestation and IgE recognition patterns to kiwifruit allergens. J Allergy Clin Immunol, 2013. 131(1): p. 164-71.
  8. Oberhuber, C., et al., Characterization of Bet v 1-related allergens from kiwifruit relevant for patients with combined kiwifruit and birch pollen allergy. Mol Nutr Food Res, 2008. 52 Suppl 2: p. S230-40.
  9. Matricardi, P.M., et al., EAACI Molecular Allergology User's Guide. Pediatr Allergy Immunol, 2016. 27 Suppl 23: p. 1-250.
  10. Bublin, M., et al., Component-resolved diagnosis of kiwifruit allergy with purified natural and recombinant kiwifruit allergens. J Allergy Clin Immunol, 2010. 125(3): p. 687-94, 694 e1.
  11. Westman, M., et al., Early childhood IgE reactivity to pathogenesis-related class 10 proteins predicts allergic rhinitis in adolescence. J Allergy Clin Immunol, 2015. 135(5): p. 1199-206 e1-11.
  12. Chruszcz, M., et al., Structural and bioinformatic analysis of the kiwifruit allergen Act d 11, a member of the family of ripening-related proteins. Mol Immunol, 2013. 56(4): p. 794-803.
  13. Zeindl, R. and M. Tollinger, NMR resonance assignments of the PR-10 allergens Act c 8 and Act d 8 from golden and green kiwifruit. Biomol NMR Assign, 2021. 15(2): p. 367-371.
  14. IUIS/WHO. IUIS/WHO Act d 8. 2022  2022 April 26]; Available from: http://allergen.org/viewallergen.php?aid=14.
  15. Scala, E., et al., Molecular Recognition Profiles and Clinical Patterns of PR-10 Sensitization in a Birch-Free Mediterranean Area. Int Arch Allergy Immunol, 2017. 173(3): p. 138-146.