Whole Allergen
g7 Common reed
g7 Common reed Scientific Information
Type:
Whole Allergen
Display Name:
Common reed
Allergen code:
g7
Family:
Poaceae (Gramineae)
Species:
Phragmites communis, P. vulgaris, Arundo phragmites, A. vulgaris
Route of Exposure:
Inhalation
Latin Name:
Phragmites communis
Other Names:
Ditch reed, giant reed, reed grass
Synonyms:
Phragmites australis, P. vulgaris, Arundo phragmites, Arundo vulgaris
Summary
The long-lived perennial common reed (Phragmites australis, previously known as P. communis) is one of the most widely distributed flowering plants and can be found in stationary or slow-moving aquatic habitats throughout the world. Flowering occurs in late summer in Europe, and the pollen can induce respiratory and cutaneous symptoms in sensitized individuals, however few studies have been reported to date. Variable degrees of cross-reactivity have been reported between the common reed and grasses of other subfamilies within the Poaceae.
Allergen
Nature
The common reed can be found in stationary or slow-moving waters in habitats ranging from boreal to tropical, and as a lowland or upland marginal or bankside species along many watercourses including artificial agricultural channels, ponds, ditches and salt meadows (1, 2).
The common reed grows rapidly up to 4 m (occasionally 6 m) high with roots that can reach 4 m deep, and in suitable habitats will out-compete virtually all other plant species to form a totally dominant stand (1, 2). Vegetative reproduction is mainly via aggressive spread of rhizomes and stolons through the sediment, however all fully developed shoots have the potential to flower, and cross-pollination by wind is the main vector for sexual reproduction (1, 2). Flowering occurs between late August to early September in Europe, with some variation in timing due to genetic and environmental factors (3). In South Africa, grass pollens including that of common reed were recovered from a spore trap throughout the year but peaked during October to December (4).
Taxonomy 
Taxonomic tree of Phragmites australis (common reed) (1)
Domain
Eukaryota
Kingdom
Plantae
Phylum
Spermatophyta
Subphylum
Angiospermae
Class
Monocotyledonae
Order
Cyperales
Family
Poaceae
Genus
Phragmites
The common reed is a phenotypically, genetically and cytologically variable plant, long known by its synonym P. communis, but now universally known as P. australis (also P. vulgaris) (1). P. australis may be considered to form a species complex as infraspecific taxa within P. australis are currently unresolved (2).
Environmental Characteristics
Worldwide distribution
One of the most widely distributed of all flowering plants, the long-lived perennial common reed is highly adaptable to freshwater and brackish conditions and considered an extremely important native species of natural wetlands and mudflats in Eurasia, North America, Africa and Australia (1, 2).
Regarded as one of the most severe causes of emergent weed problems in freshwater systems in Europe and many other places worldwide, the common reed can block channel water flow and invade a wide range of crops adjacent to drainage or irrigation canals such as cotton and rice (1). However, common reed may also be deliberately planted as part of wastewater management systems, to prevent soil erosion and to stabilize riverbanks, and is widely cultivated for its many uses in thatching, traditional crafts and medicine, pulp production, human consumption and fodder for livestock (1).
Route of Exposure
Main 
Pollen from common reed is an aeroallergen that can elicit respiratory and cutaneous symptoms in sensitized individuals (5-7).
Clinical Relevance
IgE-mediated reactions
Anecdotal evidence suggests that pollen from common reed can induce asthma, allergic rhinitis, allergic conjunctivitis and cutaneous symptoms in sensitised individuals; however, few studies have been reported to date (5-7).
A study in Spain reported that 90% (27/30) of patients with respiratory symptoms during grass pollen season were sensitized to common reed (cited by the authors as P. communis) (6). Of the 31 patients initially recruited into this study (one patient was excluded), all had rhinitis, 20 had conjunctivitis, 6 had asthma and 6 had cutaneous symptoms (6). 
Molecular Aspects
Allergenic molecules
The following allergens have been characterized from Phragmites australis pollen (5):      
Allergen
Type
Group
Molecular Weight
Phr a 1
Expansin
1
30
Phr a 4
Berberine bridge enzyme
4
60
Phr a 5
Ribonuclease
5
35
Phr a 12
Profilin
n/d
14
Phr a 13
Polygalacturonase
13
-
Biomarkers of severity
Group 13 allergens specifically occur in pollen of the major grass subfamilies, and Group 13-specific IgE antibodies can be considered immunological markers for genuine grass pollen sensitization (8).
Cross-reactivity
A certain degree of cross-reactivity among members of the Poaecea family may be expected as most species of grass pollen contain at least the Group 1, Group 5 and Group 13 grass allergens (8, 9). However, the extent of cross-reactivity between common reed and other grass species may depend on their taxonomic relationship, with significantly higher average inhibition of IgE binding likely for pollen extracts of grasses within the same group, and lower levels of inhibition seen for grasses that are more distantly related (9, 10). For example, Duffort et al. (2008) demonstrated that antigentic similarity among Group 1 allergens is very high among grasses in the Pooideae subfamily, but lower among grasses from other subfamilies such as Arundinoideae (common reed, P. communis) and Chloridoideae (Bermuda grass, C. dactylon) (11).
Timothy grass pollen Phl p2 cross-reacted weakly with Group 2 allergens from cultivated oat and common reed pollens (12).
Pollen from 17 grass species including common reed (cited by the authors as P. communis) was used to test IgE reactivity of sera from 800 individuals suspected of an inhalant allergy in North America and Europe (7). Within this cohort, specific IgE antibodies against one or more grass species were detected in about a quarter (26.1%, 209/800) of sera, however IgE responses to P. communis were lower - by a factor of approximately 5 - than those to species in the Pooideae subfamily such as Kentucky bluegrass (Poa pratensis), creeping fescue (Festuca rubra), timothy grass (Phleum pratense), orchard grass (Dactylis glomerata) and ryegrass (Lolium perenne) (7).
A study in Spain reported that 90% (27/30) of patients with respiratory symptoms such as rhinitis and/or asthma during grass pollen season and specific IgE to a grass mixture extracted from grasses from the Pooideae subfamily were also sensitized to common reed (cited by the authors as P. communis), and 80% (24/30) were sensitized to Bermuda grass (Cynodon dactylon) (6). Of the 31 patients initially recruited into this study (one patient was excluded), all had rhinitis, 20 had conjunctivitis, 6 had asthma and 6 had cutaneous symptoms (6). 
Compiled By
Author: RubyDuke Communications
Reviewer: Dr. Christian  Fischer 
Last reviewed:May 2022
References
  1. CABI. Phragmites australis (common reed) 2021 [cited 2021 3.12.21]. Available from: https://www.cabi.org/isc/datasheet/40514
  2. Packer JG, Meyerson LA, Skálová H, Pyšek P, Kueffer C. Biological Flora of the British Isles: Phragmites australis. Journal of Ecology. 2017;105(4):1123-62.
  3. McKee J, Richards AJ. Variation in seed production and germinability in common reed (Phragmites australis) in Britain and France with respect to climate. New Phytologist. 1996;133(2):233-43.
  4. Potter PC, Berman D, Toerien A, Malherbe D, Weinberg EG. Clinical significance of aero-allergen identification in the western Cape. S Afr Med J. 1991;79(2):80-4.
  5. Allergome. Phr a (common reed) Wallingford, UK2021 [cited 2021 3.12.21]. Available from: http://www.allergome.org/script/dettaglio.php?id_molecule=1927.
  6. López-Matas MA, Moya R, Cardona V, Valero A, Gaig P, Malet A, et al. Relevance of Allergenic Sensitization to Cynodon dactylon and Phragmites communis: Cross-reactivity With Pooideae Grasses. J Investig Allergol Clin Immunol. 2016;26(5):295-303.
  7. van Ree R, van Leeuwen WA, Aalberse RC. How far can we simplify in vitro diagnostics for grass pollen allergy?: A study with 17 whole pollen extracts and purified natural and recombinant major allergens. J Allergy Clin Immunol. 1998;102(2):184-90.
  8. Grote M, Swoboda I, Valenta R, Reichelt R. Group 13 allergens as environmental and immunological markers for grass pollen allergy: studies by immunogold field emission scanning and transmission electron microscopy. Int Arch Allergy Immunol. 2005;136(4):303-10.
  9. Van Ree R, Driessen MN, Van Leeuwen WA, Stapel SO, Aalberse RC. Variability of crossreactivity of IgE antibodies to group I and V allergens in eight grass pollen species. Clin Exp Allergy. 1992;22(6):611-7.
  10. Niederberger V, Laffer S, Fröschl R, Kraft D, Rumpold H, Kapiotis S, et al. IgE antibodies to recombinant pollen allergens (Phl p 1, Phl p 2, Phl p 5, and Bet v 2) account for a high percentage of grass pollen-specific IgE. J Allergy Clin Immunol. 1998;101(2 Pt 1):258-64.
  11. Duffort O, Quintana J, Ipsen H, Barber D, Polo F. Antigenic similarity among group 1 allergens from grasses and quantitation ELISA using monoclonal antibodies to Phl p 1. Int Arch Allergy Immunol. 2008;145(4):283-90.
  12. Flicker S, Steinberger P, Norderhaug L, Sperr WR, Majlesi Y, Valent P, et al. Conversion of grass pollen allergen-specific human IgE into a protective IgG(1) antibody. Eur J Immunol. 2002;32(8):2156-62