Whole Allergen
m209 Penicillium glabrum
m209 Penicillium glabrum Scientific Information
Type:
Whole Allergen
Display Name:
Penicillium glabrum
Allergen code:
m209
Species:
glabrum
Latin Name:
Penicillium glabrum
WHO/ICD-11 code:
XN1ZF
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Summary
Penicillium glabrum is a saprophyte fungus part of the Penicillium genus, one of the four most common fungal causes of allergy in humans. P. glabrum is associated with suberosis, an occupational hypersensitivity associated with cork production. It is also a mold commonly found in homes. Although no P. glabrum specific allergen is recognized, two protein components of 12–13.5 kDa and 33 kDa are significant in the pathogenesis of suberosis. 
Allergen
Nature
Penicillium spp. are commonly isolated in indoor and outdoor environments. The molds found on fruit and vegetables are typically blue-green and belong to the Penicillium genus; they are used commercially in the production of blue cheese (1). Penicillium is also one of the four most common fungal genera associated with allergy in human patients (2). They are saprophytes that produce spores prolifically and can cause both allergic asthma and occupational pneumonitis in humans (3).
Penicillium glabrum is the main causative agent of suberosis, an occupational allergen associated with Hypersensitive Pneumonitis (HP) in workers in the cork production industry (3). P. glabrum forms orange, orange-brown and occasionally cream colonies. Microscopic features of the species include a primary monoverticillate morphology, with vesiculate conidiophores and conidia that or smooth or slightly rugose (4). 
Taxonomy
Taxonomic tree of Penicillium glabrum (1)
Domain
Eukaryote
Kingdom
Fungi
Sub Kingdom
Dikarya
Phylum
Ascomycota
Subphylum
Pezizomycotina
Class
Eurotiomycetes
Subclass
Eurotiomycetidae
Order
Eurotiales
Family
Trichomaceae
Genus
Globosum
Epidemiology
Worldwide distribution 
A study from the USA found that 8.2% of a cohort comprising of children with asthma were sensitized to Penicillium spp. (5). Elsewhere, sensitization to a Penicillium allergen, Pen Ch, was relatively low amongst patients with atopic dermatitis (6). Another study found that 100% (n=16) of workers with a history of occupational exposure to cork and who had either HP or asthma were sensitized to P. glabrum specific allergens. However, the immunological response reported was a specific IgG response, with no IgE response found (3). In Portugal, it is estimated that 9-19% of cork workers may develop suberosis (7). 
Environmental Characteristics
Living environment
Penicillium spp., as xerophilic fungi, are characterized as indoor allergens (6) and are one of the three most common indoor airborne fungi (1). One study has found Penicillium spp. present in 33 % of homes (n=1,233) in the North Eastern United States (5). Furthermore, a survey of homes in Eastern France found that P. glabrum was the second most prevalent Penicillium species (22.9%) isolated (8). 
Route of Exposure
Main
Exposure and sensitization to P. glabrum allergens are most commonly achieved by inhaling fungal fragments (7). 
Clinical Relevance
Penicillium spp. are considered by the institute of medicine to be causative agents of allergic rhinitis in children and adults, as well as asthma children and occupational asthma in adults (8).
In contrast to other allergenic molds, a study in the USA found that increased prevalence of Penicillium correlated with increased risk of clinically apparent hypersensitivity (incident wheeze and persistent cough) (5). Children who were both sensitized to Penicillium spp. and who lived in a household where Penicillium spp. were detected, even at low levels, were twice as likely to experience a higher asthma severity score than the cohort who were either not sensitized or exposed. This contrasts with data for another fungal genus, Cladosporium, which showed sensitivity and exposure status were not associated with clinical outcomes (5).
HP is a lung disease characterized by lymphocytic – histiocytic inflammation of the alveoli, bronchioles and pulmonary interstitium, mainly through inhalation of organic substances. HP is probably underdiagnosed (9). Suberosis, of which P. glabrum is the primary causative agent (3), is a form of HP associated with repeated exposure to moldy cork dust in workplaces where this material is used in industrial processing. 
Molecular Aspects
Allergenic molecules
Although the WHO-IUIS Allergen Nomenclature Sub-Committee recognizes multiple allergens from Penicillium species, their role in the pathogenesis of Penicillium hypersensitivity is yet to be described. The major Penicillium allergens are proteases (alkaline and vacuolar serine proteases) and are referred to as group 14 and group 18 allergens due to their high sequence homology with the Aspergillus fumigatus allergens Asp f 14 and Asp f 18. Furthermore, there are no approved allergens specific to P. glabrum (1).
One study, which aimed to characterize the P.glabrum allergen, using sera from patients with suberosis or asthma, found two P. glabrum proteins that could be candidate allergens. The components (12–13.5 kDa and 33 kDa) were significantly associated with clinical HP. Although the study did not demonstrate IgE binding of the 33 kDa component, IgE binding has been shown with components of 33 kDa in other Penicillium spp. (3).
Cross-reactivity
Mono-sensitization to P. glabrum is unlikely as the Penicillium specific allergens described are characteristically cross-reactive (1).
Compiled By
Author: RubyDuke Communications
Reviewer: Dr. Christian  Fischer
Last reviewed:March 2022
References
  1. Fukutomi Y, Taniguchi M. Sensitization to fungal allergens: Resolved and unresolved issues. Allergol Int. 2015;64(4):321-31.
  2. Twaroch TE, Curin M, Valenta R, Swoboda I. Mold allergens in respiratory allergy: from structure to therapy. Allergy Asthma Immunol Res. 2015;7(3):205-20.
  3. Winck JC, Delgado L, Murta R, Vanzeller M, Marques JA. Cork workers' occupational asthma: lack of association with allergic sensitisation to fungi of the work environment. Int Arch Occup Environ Health. 2004;77(4):296-300.
  4. Barreto MC, Houbraken J, Samson RA, Frisvad JC, San-Romão MV. Taxonomic studies of the Penicillium glabrum complex and the description of a new species P. subericola. Fungal Diversity. 2011;49(1):23-33.
  5. Gent JF, Kezik JM, Hill ME, Tsai E, Li DW, Leaderer BP. Household mold and dust allergens: exposure, sensitization and childhood asthma morbidity. Environ Res. 2012;118:86-93.
  6. Celakovska J, Vankova R, Bukac J, Cermakova E, Andrys C, Krejsek J. Atopic Dermatitis and Sensitisation to Molecular Components of Alternaria, Cladosporium, Penicillium, Aspergillus, and Malassezia-Results of Allergy Explorer ALEX 2. J Fungi (Basel). 2021;7(3).
  7. Nogueira R, Melo N, Novais EBH, Martins N, Delgado L, Morais A, et al. Hypersensitivity pneumonitis: Antigen diversity and disease implications. Pulmonology. 2019;25(2):97-108.
  8. Reboux G, Rocchi S, Vacheyrou M, Millon L. Identifying indoor air Penicillium species: a challenge for allergic patients. Journal of Medical Microbiology. 2019;68(5):812-21.
  9. Morell F, Cruz MJ, Gómez FP, Rodriguez-Jerez F, Xaubet A, Muñoz X. Chacinero's lung - hypersensitivity pneumonitis due to dry sausage dust. Scand J Work Environ Health. 2011;37(4):349-56