Whole Allergen
p4 Anisakis
p4 Anisakis Scientific Information
Type:
Whole Allergen
Display Name:
Anisakis
Allergen code:
p4
Family:
Anisakidae
Species:
Anisakis spp.
Route of Exposure:
Ingestion
Latin Name:
Anisakis spp
WHO/ICD-11 code:
XN574
(ICD-11 is currently under implementation by WHO and the ICD-11 codes displayed in the encyclopedia may not yet be available in all countries)
Summary
Anisakis spp. are parasitic nematodes of many marine species that are globally distributed and can infect humans via consumption of raw, undercooked, or processed food products. Anisakis infection causes ‘anisakiasis’, a clinical disease with nonspecific clinical manifestations including epigastric pain, nausea, vomiting, abdominal distention with intense pain, and occasionally hypersensitivity or anaphylaxis. Anisakiasis is a common form of food poisoning with tens of thousands of cases reported worldwide. Sensitized individuals can also develop Anisakis-induced asthma, rhinoconjunctivitis, dermatitis, and secondary gingivostomatitis following exposure to proteins of dead larvae. Approximately 26 allergens with a variety of biological functions have been characterized from Anisakis spp. to date, many of which persist and are highly resistant to heat and digestive enzymes. Major allergens include Ani s 1, Ani s 7, Ani s 12 and Ani s 14, while minor allergens are considered to be Ani s 4, Ani s 5, Ani s 6, Ani s 8, Ani s 9, Ani s 10, and Ani s 11. Ani s 2 (paramyosin) and Ani s 3 (tropomyosin) are considered pan-allergens with low specificity.
Allergen
Nature
Anisakis is a genus of nematodes with approximately 12 species that infect a variety of marine fish and cephalopods as intermediate hosts at different stages in their life cycle (1-4). The Anisakidae family is characteristically adaptable, radiating through trophic links of marine ecosystems to infest different hosts from merozooplankton, up to top predators such as pinnipeds and cetaceans (2, 4).
Anisakis has four larval stages, L1‒L4, with female L4 capable of producing 1.5 million eggs (2). Third-stage larvae (L3) of Anisakis can infect humans who eat raw, undercooked, or processed (e.g. smoked, salted, brined) parasitized fish and cephalopod products (1, 4-7).
Larvae are 1‒3 cm in length (5). Visual identification of Anisakis is complicated by various factors: the various life cycle stages, morphological characteristics shared by different genera such as relative size and shape, and damage or fragmentation of the parasite on removal (2, 8). Precise species identification relies on molecular evidence such as genetic sequencing, and the majority of human reports identify the parasite as “Anisakis larval type” (2, 3, 8). 
Taxonomy 
Taxonomic tree of Anisakis spp. (9)
Domain
Eukaryota
Kingdom
Animalia
Phylum
Nematoda
Class
Chromadorea
Family
Anisakidae
Genus
Anisakis
Environmental Characteristics
Living environment
Depending on the stage of the life cycle, Anisakis can be found within a variety of marine host species or free-living in marine waters (2, 4, 5, 7).
Anisakis stage L4 produce eggs in the intestines of cetaceans, their definitive hosts, that pass into the water in feces and develop to larval stages L1 and L2 (2, 7). Intermediate marine hosts then acquire Anisakis by preying on infected animals or ingesting larvae from the water. Within the intermediate host, Anisakis larvae develop to stage L3 which can penetrate the stomach wall, migrate into the visceral cavity, settle on the external surface of organs such as the liver, gonads, and mesentery, and be encapsulated into the tissues (2, 7, 10). The life cycle of the parasite is completed when the intermediate host is ingested by marine mammals or birds, and the adult form (L4) can develop (2, 7).
Humans can be accidental hosts of Anisakis by consuming the intermediate marine host species, however, parasitic reproduction does not take place as larval L3 cannot develop to adult L4 in intermediate hosts or humans (2, 7). In humans, Anisakis larvae L3 may burrow into gastric mucosa and submucosa to avoid the highly acidic stomach environment (1, 2, 5). Larvae usually die within a few days to weeks after human consumption and are broken down in approximately eight weeks, during which time larval remains are surrounded by edema, necrosis, and cellular inflammation, deposition of fibrotic tissues and formation of foreign body giant cells and lymphocytes, and ultimately a granuloma (2, 5).
Worldwide distribution 
Anisakis spp. is located mainly in Europe, Asia, Australia, North America, and South America (2). However, because a variety of highly-mobile marine cetaceans and pinniped species serve as definitive hosts for Anisakis, the infectious L3 development stage can be commonly found in fish, cephalopods, marine birds, and other animals worldwide (2, 8). A recent systematic review reported allergic anisakiasis hot spots in the north and northeast Atlantic Ocean, southwest of USA, west of Mexico, south of Chile, east of Argentina, Norway, UK, and west of Iceland (confidence 99%) (8).
Route of Exposure
Main 
Ingestion of raw, undercooked, or processed (e.g. smoked, salted, brined, or canned) parasitized fish and cephalopod products that contain the Anisakis third-stage larvae (L3) (1, 2, 5, 7, 10, 11).
Clinical Relevance
Anisakis infection in humans causes ‘anisakiasis’, a clinical disease with nonspecific clinical manifestations including epigastric pain, nausea, vomiting, abdominal distention with intense pain, and occasionally hypersensitivity or anaphylaxis (2, 5, 7). Pathogenicity of anisakiasis results from direct tissue damage and/or an allergic response to the release of potent proteolytic enzymes and other metabolic products from the parasite (2, 4). Anisakiasis is a common form of food poisoning (2, 4, 13). Tens of thousands of cases of anisakiasis reported from Europe, Asia, and other parts of the world can be grouped into four common symptomatic clinical manifestations: gastric, intestinal, ectopic (extra-intestinal), and allergic (4, 5, 8). Sensitization rates to Anisakis have increased worldwide with a significant impact on healthcare systems (14).
Allergic anisakiasis is characterized by angioedema, urticaria, hypersensitivity syndrome, or severe anaphylactic reaction, with immediate onset in IgE-mediated severe cases, or between 60‒120 minutes after digestion of infected food (4, 8). Urticaria has been reported in 60‒70% of cases where there is a gastric presentation (2). Evidence suggests that humans previously sensitized to Anisakis can experience escalation of symptoms associated with allergic anisakiasis upon subsequent challenge, which can be serious or life-threatening. A recent systematic review reported the highest global rates of allergic anisakiasis in Portugal and Norway, with a prevalence rate of 18.45% to 22.50%, followed by Spain, Sweden, and Japan (8).
In most cases, initial sensitization and subsequent gastroallergic reactions are caused by exposure to live Anisakis larvae, however, sensitized individuals can also develop Anisakis-induced asthma, rhinoconjunctivitis, dermatitis, and secondary gingivostomatitis following exposure to proteins of dead larvae (2, 4, 5, 8). Occupational exposure to parasitized fish can elicit allergic reactions including bronchial hyperreactivity and dermatitis (7).
Polymerase chain reaction (PCR) tests on intestinal biopsies from anisakiasis patients indicate a Th2 type immune response as T-cell receptor and Th2 cytokines IL-4 and IL-5 were detected, when IFN-gamma or IL-2 was not detected.Th2 cytokines are responsible for symptoms of gastroallergic anisakiasis as well as Anisakis allergy, which can manifest as asthma, rhinoconjunctivitis, urticaria, and atopic dermatitis, primarily driven by mediators released from mast cells (4, 5, 12). Gastroallergic anisakiasis can potentiate non-steroidal anti-inflammatory drug (NSAID)-induced upper gastrointestinal bleeding, and is associated with autoimmune disease, nontolerance of oral antigens, increasing susceptibility to secondary infections, and decreasing vaccine efficacy (4, 12). Anisakis-induced granulomas have been mistaken for tumors and may persist for some time, leading to symptoms of chronic anisakiasis (5). Ectopic anisakiasis is less common (5) but also has been associated with anaphylaxis in a case study of a person with scrotal localization, (15).
Diagnostics Sensitization
Diagnosis of allergic anisakiasis is usually based on serology tests that are non-specific (8, 12). The presence of specific IgE or a positive skin prick test alone does not indicate allergy, which can only be assessed by a clinical history, but sensitization (5). Of note, specific IgE detection by ImmunoCAP assay can overestimate the number of sensitized subjects (4).
Molecular Aspects
Allergenic molecules
The following allergens and their molecular epitopes have been characterized from Anisakis pegreffi (Ani pe) and Anisakis simplex (Ani s) (16, 17):
Name
Type
Mass (kDa)
Ani pe 1
Serine protease inhibitor
-
Ani pe 2
Paramyosin
-
Ani pe 12
Unknown function
-
Ani pe 13
Hemoglobin
35 (18)
Ani s 1, Ani s 1.0101
Serine protease inhibitor
24
Ani s 2, Ani s 2.0101
Paramyosin
97
Ani s 3, Ani s 3.0101,
Ani s 3.0102
Tropomyosin
-
Ani s 4, Ani s 4.0101
Cystatin
9
Ani s 5, Ani s 5.0101
SXP/RAL-2 protein
15
Ani s 6, Ani s 6.0101
Serine protease inhibitor
-
Ani s 7, Ani s 7.0101
Unknown function
139
Ani s 8, Ani s 8.0101
SXP/RAL-2 proteins
15
Ani s 9, Ani s 9.0101
SXP/RAL-2 proteins
14
Ani s 10, Ani s 10.0101
Unknown function
21
Ani s 11, Ani s 11.0101,
Ani s 11.0201
Unknown function
-
Ani s 12, Ani s 12.0101
Unknown function
-
Ani s 13, Ani s 13.0101
Hemoglobin
37
Ani s 14, Ani s 14.0101
Unknown function
23.5  (19)
Ani s 24kD
Unknown function
-
Ani s CCOS3
Cytochrome C oxidase
-
Ani s Cytochrome B
Cytochrome B protein
-
Ani s Enolase
Enolase
48
Ani s FBPP
Fructose-1,6-biphosphatase
-
Ani s NADHDS4L
NADH dehydrogenase
-
Ani s NARaS
Nicotinic acetylcholine receptor protein
-
Ani s PEPB
Phosphatidylethanolamine-binding protein
-
Ani s Troponin
Troponin
-
Approximately 26 allergens with a variety of biological functions have been characterized from Anisakis spp. to date, many of which persist and are highly resistant to heat and digestive enzymes (2, 7). There is some evidence that Anisakis differentially expresses certain proteins including allergens, up-regulating or down-regulating in response to abiotic conditions such as temperature, anatomical infection site in the intermediate host, and host-specific immune evasion mechanisms (7, 10). Differences in IgE reactivity in sera of patients infected with Anisakis could reflect interindividual variations in immunological response, a different frequency of exposure, or patient selection bias when comparing different populations (12).
Major allergens include Ani s 1, Ani s 7, Ani s 12 and Ani s 14, while minor allergens are considered to be Ani s 4, Ani s 5, Ani s 6, Ani s 8, Ani s 9, Ani s 10, and Ani s 11 (2, 12, 19). Ani s 2 (paramyosin) and Ani s 3 (tropomyosin) are considered pan-allergens with low specificity (12).
The serum protein Ani s 1 is identified in 87% of patients who develop a clinical picture after ingesting infected fish (2). Ani s 7 is recognized by more than 90% of patients, and IgE antibodies to Ani s 7 are consistently high in patients with acute Anisakis infection (gastroallergic anisakiosis) or Anisakis infection associated with chronic urticaria (2, 12). Recombinant Ani s 14 was shown to be IgE-reactive to 14 (56%) of 26 sera from Anisakis-allergic patients (19).
Ani s 4, detected in 27‒30% of allergic patients, is resistant to autoclaving and pepsin digestion and, along with other allergens resistant to heat treatment or enzyme digestion (e.g. Ani s 1 and allergens from the SXP/Ral family such as Ani s 5, Ani s 8, and Ani s 9), could therefore be clinically relevant for exposure to parasite-containing fishery products even after processing for consumption (1, 2, 11, 20). Ani s 11 is also heat-stable after boiling for 30 minutes, resistant to pepsin digestion for 120 minutes, and detected by 78% of 37 Anisakis-allergic patients, with 13.5% of patients only detecting the recombinant Ani s 11-like allergen (21).
Nematode hemoglobins have a high oxygen affinity and may represent an important component of parasite adaptation to co-existence within the host (10). Rats inoculated with live A. simplex larvae demonstrated strong and prolonged IgE and IgG immunoreactions to Ani s 13 hemoglobin (18), while a small study in humans demonstrated that native Ani s 13 was detected by 72.1% of 43 Anisakis-sensitized individuals (22). 
Cross-reactivity
Allergens from Anisakis spp. have been reported to cross-react with molecules from mites, crustaceans, insects, mollusks, and other nematode parasites (7, 23, 24). In a small study, a total of 44% of 25 Anisakis-allergic patients had specific IgE to Vespula spp. wasp venom and positivity to at least one of the Hymenoptera allergens was detected in 16% of individuals (25). 
Compiled By
Author: RubyDuke Communications
Reviewer: Dr.Christian Fischer
Last reviewed: June 2022
References
  1. Polimeno L, Lisanti MT, Rossini M, Giacovazzo E, Polimeno L, Debellis L, et al. Anisakis Allergy: Is Aquacultured Fish a Safe and Alternative Food to Wild-Capture Fisheries for Anisakis simplex-Sensitized Patients? Biology. 2021;10(2):106.
  2. Ángeles-Hernández JC, Gómez-de Anda FR, Reyes-Rodríguez NE, Vega-Sánchez V, García-Reyna PB, Campos-Montiel RG, et al. Genera and Species of the Anisakidae Family and Their Geographical Distribution. Animals (Basel). 2020;10(12):2374.
  3. S CA, Robertson L, Ciordia S, Sánchez-Alonso I, Careche M, Carballeda-Sanguiao N, et al. Quantitative Proteomics Comparison of Total Expressed Proteomes of Anisakis simplex Sensu Stricto, A. pegreffii, and Their Hybrid Genotype. Genes (Basel). 2020;11(8).
  4. Audicana MT, Kennedy MW. Anisakis simplex: from obscure infectious worm to inducer of immune hypersensitivity. Clinical microbiology reviews. 2008;21(2):360-79.
  5. Nieuwenhuizen NE. Anisakis - immunology of a foodborne parasitosis. Parasite Immunol. 2016;38(9):548-57.
  6. Suzuki J, Murata R, Kodo Y. Current Status of Anisakiasis and Anisakis Larvae in Tokyo, Japan. Food Saf (Tokyo). 2021;9(4):89-100.
  7. Mehrdana F, Buchmann K. Excretory/secretory products of anisakid nematodes: biological and pathological roles. Acta Veterinaria Scandinavica. 2017;59(1):42.
  8. Rahmati AR, Kiani B, Afshari A, Moghaddas E, Williams M, Shamsi S. World-wide prevalence of Anisakis larvae in fish and its relationship to human allergic anisakiasis: a systematic review. Parasitol Res. 2020;119(11):3585-94.
  9. ITIS. Anisakis 2021 [cited 2022 27.01.22]. Available from: https://www.itis.gov/servlet/SingleRpt/SingleRpt?search_topic=TSN&search_value=63864#null.
  10. Palomba M, Cipriani P, Giulietti L, Levsen A, Nascetti G, Mattiucci S. Differences in Gene Expression Profiles of Seven Target Proteins in Third-Stage Larvae of Anisakis simplex (Sensu Stricto) by Sites of Infection in Blue Whiting (Micromesistius poutassou). Genes (Basel). 2020;11(5).
  11. Kochanowski M, Różycki M, Dąbrowska J, Bełcik A, Karamon J, Sroka J, et al. Proteomic and Bioinformatic Investigations of Heat-Treated Anisakis simplex Third-Stage Larvae. Biomolecules. 2020;10(7).
  12. Ubeira FM. Travelling with Anisakis allergens. Int Arch Allergy Immunol. 2014;163(4):243-4.
  13. Suzuki T, Kusano K, Kondo N, Nishikawa K, Kuge T, Ohno N. Biological Activity of High-Purity β-1,3-1,6-Glucan Derived from the Black Yeast Aureobasidium pullulans: A Literature Review. Nutrients. 2021;13(1).
  14. Mazzucco W, Raia DD, Marotta C, Costa A, Ferrantelli V, Vitale F, et al. Anisakis sensitization in different population groups and public health impact: A systematic review. PLOS ONE. 2018;13(9):e0203671.
  15. Centonze A, Capillo S, Mazzei A, Salerno D, Sinopoli D, Prosperi Porta I, et al. Acute scrotum in a 8-year-old Italian child caused by extraintestinal anisakiasis in a seaside area. Allergy. 2021;76(5):1601-2.
  16. allergome.org. Ani S 2021 [cited 2022 27.01.22]. Available from: https://www.allergome.org/script/dettaglio.php?id_molecule=1716.
  17. allergome.org. Ani pe 2021 [cited 2022 27.01.22]. Available from: https://www.allergome.org/script/dettaglio.php?id_molecule=2918.
  18. Abe N, Teramoto I. Anisakis haemoglobin is a main antigen inducing strong and prolonged immunoreactions in rats. Parasitol Res. 2017;116(7):2035-9.
  19. Kobayashi Y, Kakemoto S, Shimakura K, Shiomi K. Molecular Cloning and Expression of a New Major Allergen, Ani s 14, from Anisakis simplex. Shokuhin Eiseigaku Zasshi. 2015;56(5):194-9.
  20. Rodriguez-Mahillo AI, Gonzalez-Muñoz M, Gomez-Aguado F, Rodriguez-Perez R, Corcuera MT, Caballero ML, et al. Cloning and characterisation of the Anisakis simplex allergen Ani s 4 as a cysteine-protease inhibitor. Int J Parasitol. 2007;37(8-9):907-17.
  21. Carballeda-Sangiao N, Rodríguez-Mahillo AI, Careche M, Navas A, Caballero T, Dominguez-Ortega J, et al. Ani s 11-Like Protein Is a Pepsin- and Heat-Resistant Major Allergen of Anisakis spp. and a Valuable Tool for Anisakis Allergy Component-Resolved Diagnosis. Int Arch Allergy Immunol. 2016;169(2):108-12.
  22. González-Fernández J, Rivas L, Luque-Ortega JR, Núñez-Ramírez R, Campioli P, Gárate T, et al. Recombinant vs native Anisakis haemoglobin (Ani s 13): Its appraisal as a new gold standard for the diagnosis of allergy. Experimental parasitology. 2017;181:119-29.
  23. Verga MC, Pastorino R, Casani A, Inturrisi F, de Waure C, Pugliese A, et al. Prevalence, molecular characterization, and clinical relevance of sensitization to Anisakis simplex in children with sensitization and/or allergy to Dermatophagoides pteronyssinus. Eur Ann Allergy Clin Immunol. 2017;49(6):270-5.
  24. Fernández-Caldas E, Quirce S, Marañó F, Gómez MLD, Botella HG, Román RL. Allergenic cross-reactivity between third stage larvae of <strong><em>Hysterothylacium aduncum</em></strong> and <strong><em>Anisakis simplex</em></strong>. Journal of Allergy and Clinical Immunology. 1998;101(4):554-5.
  25. Rodriguez-Perez R, Crespo JF, Rodríguez J, Salcedo G. Profilin is a relevant melon allergen susceptible to pepsin digestion in patients with oral allergy syndrome. J Allergy Clin Immunol. 2003;111(3):634-9.