Type:
Whole Allergen
Whole Allergen
Whole Allergen
Horse chestnut
t203
Hippocastanaceae
Aesculus hippocastanum
Horse chestnut tree, Common horse chestnut
Anecdotal evidence suggests that asthma, allergic rhinitis and allergic conjunctivitis are possible following exposure to pollen from this tree; however, few specific studies have been reported to date (1-2).
Sensitisation to Horse chestnut tree pollen is common where the tree is growing. In an Austrian study, IgE antibodies to Horse chestnut pollen were found in 12.6% of urban children, compared to 1.9% of control subjects recruited from a rural area. Horse chestnut-specific IgE levels were highest in highly atopic pollen-allergic children, in particular those also sensitised to the Plane tree (1).
Contact dermatitis to the extract of Horse chestnut has been reported (3).
No allergens from this plant have yet been characterized
An extensive cross-reactivity among the different individual species of the genus could be expected (4).
Last Reviewed- April 2022
Before you proceed ...
We noticed you may be visiting a version of our website that doesn’t match your current location. Would you like to view content based on your region?
Talk with us
Interested in utilizing our solutions in your laboratory or healthcare practice?
This form is dedicated to healthcare and laboratory professionals who work within medical laboratories, private practices, health systems, and the like.
By clicking "continue with form" you are confirming that you work within a healthcare or laboratory space.
Not a lab professional or clinician but interested in diagnostic testing for allergies?
Visit Allergy InsiderChoose your preferred language
Please note: By selecting a different language, you are choosing to view another site. Product availability and indications may vary from what is approved in your region.