Type:
Whole Allergen
Whole Allergen
Whole Allergen
Paloverde
t219
Fabaceae
Cercidium floridum
Parkinsonia florida
Blue paloverde leaves and stems contain cyanogenic glycosides, alkaloids, and cinnamic phenolic acid.
Blue paloverde is a spiny, small, deciduous tree with multiple stems, distributed throughout the Sonora Desert in the USA. Its range extends from central and southwestern Arizona into southeastern California.
Blue paloverde grows about 10 m tall. It has pinnately compound leaves. Inflorescences of Blue paloverde are 4 to 12 cm long, with 1 or more flowers. Flower and leaf production of Blue paloverde vary according to the amounts of precipitation received. Blue paloverde remains leafless throughout most of the year. Leaves are produced between mid-July and late November, depending on the summer rains. The fruits are flat legumes, each containing 1 to 8 flat seeds.
Blue paloverde typically flowers sporadically after rains from late March to May, but flowering may extend into July. Populations of Blue paloverde also may bloom from August to October
Blue paloverde occurs in arid and semiarid climates characterized by high summer temperatures and highly variable rainfall. Blue paloverde grows predominantly in washes and beside culverts and bridges, but is also found in uplands. Blue paloverde fruits have been used by Native Americans for food. The Pima and Papago in Arizona cooked young Blue paloverde fruits and seeds and ground the seeds for porridge.
No allergens from this plant have yet been characterised.
An extensive cross-reactivity among the different individual species of the genus could be expected (1).
Anecdotal evidence suggests that asthma, allergic rhinitis and allergic conjunctivitis are possible following exposure to pollen from this tree; however, no specific studies have been reported to date.
Before you proceed ...
We noticed you may be visiting a version of our website that doesn’t match your current location. Would you like to view content based on your region?
Talk with us
Interested in utilizing our solutions in your laboratory or healthcare practice?
This form is dedicated to healthcare and laboratory professionals who work within medical laboratories, private practices, health systems, and the like.
By clicking "continue with form" you are confirming that you work within a healthcare or laboratory space.
Not a lab professional or clinician but interested in diagnostic testing for allergies?
Visit Allergy InsiderChoose your preferred language
Please note: By selecting a different language, you are choosing to view another site. Product availability and indications may vary from what is approved in your region.