Whole Allergen
w82 Careless weed
w82 Careless weed Scientific Information
Type:
Whole Allergen
Display Name:
Careless weed
Allergen code:
w82
Latin Name:
Amaranthus palmeri
Environmental Characteristics
Other topics
The amaranth family comprises about 40 genera and 475 species. They are mostly weedy herbs though some genera are low or climbing shrubs. The flowers of all are characterized by extreme simplicity. Some species are wind pollinated whereas others are insect pollinated. The flowers may be monoecious or dioecious, but they are always small, often greenish or yellowish.
Careless weed is a species of flowering plant in the amaranth genus. It is native to most of the southern half of North America, and particularly, throughout the southern United States from southern California to Virginia. It has also been introduced to Europe, Australia, and other areas.
Careless weed closely resembles many other pigweed species. It is an erect summer annual that may reach 1.5-2m in height. It has one central stem from which several lateral branches arise. The leaves are alternate, glabrous (without hairs), and lance- or egg-shaped in outline. Leaf sizes are from 5 cm to 20 cm in length and 1 to 6 cm wide with prominent white veins on the under-surface. Leaves occur on relatively long petioles. (1)
The flowers are small, light-green, and inconspicuous produced in dense, compact, terminal panicles that are from 15 cm to 45 cm in length. Smaller lateral inflorescences also occur between the stem and the leaf petioles (leaf axils). Male and female flowers occur on separate plants. Each terminal panicle contains many densely packed branched spikes that have bracts that are 3 to 6 mm long. The plant blooms from June – November in the northern hemisphere. (1)
A small, dry, one seeded fruit is produced consisting of a single seeded utricle about 2 mm in size, which splits to show a glossy black to dark brown seed that is 1 to 1.2 mm long. The utricle is wrinkled when dry. (1)
Careless weed is often confused with other similar pigweed species, but differs in that no other pigweed species have terminal panicles that reach 45 cm in length, and that the terminal spike of is much smoother and narrower and less spike-like than either Common Pigweed (Redroot Pigweed) (A. retroflexus) or smooth pigweed (A. hybridus)The leaves of Careless weed are also without hairs and have prominent white veins on the under-surface unlike those of Common Pigweed. (1)
Clinical Relevance
IgE-mediated reactions
Anecdotal evidence suggests that asthma, allergic rhinitis and allergic conjunctivitis are common following exposure to pollen from Careless weed; however, few specific studies have been reported to date. (2-5)
Sensitization to Careless weed has been documented in Tucson, Arizona (5) and in Mexico. (3, 4) In a Mexican study of allergic patients, sensitization to Careless weed was demonstrated in 43.8%.(4)
In a study examining aeroallergen sensitization rates in military children in Texas undergoing skin testing for rhinitis, of 209 patients, 27% were sensitized to Common Pigweed or Careless weed. (2)
In a study in the Midwestern USA, evaluating the frequency of sensitization to cannabis pollen, found that 61% were skin prick positive for cannabis and all subjects were also skin test positive to weeds pollinating during the same period: Ragweed, Pigweed, Cocklebur, Russian thistle, Marsh elder, and Kochia. (6)
In a study among Thai patients with vernal keratoconjunctivitis, positive results of skin prick testing to Acacia, Careless weed, mold, Johnson grass and Cow's milk were significantly more common in patients with palpebral VKC. (7)
Molecular Aspects
No allergens have been characterized.
Cross-reactivity
Cross-reactivity could be expected between species of the family Amaranthaceae, and in particular the genus Amaranthus.
Although no specific information on cross-reactivity of this plant with others exists, a high degree of cross reactivity has been reported to occur between Goosefoot (Chenopodium album) and Saltwort (Salsola kali), and other species taxonomically less related members of the Amaranthaceae family like Amaranthus retroflexus. Common allergenic determinants are present in these plants. (2)
In a study using a fluorescent allergosorbent test, similar antigenic determinants were found between Short Ragweed and Giant Ragweed, Cocklebur, Lamb's Quarters, Rough Pigweed, Marsh Elder, and Goldenrod. Cocklebur and Giant Ragweed were highly potent in competitively binding to short ragweed IgE. The other pollens demonstrated lower potency of cross-reacting antigens. (3)
Compiled By
References
  1. palmeri. VTWIGPAA. http://www.ppws.vt.edu/scott/weed_id/amapa.html 2011 [cited 2011].
  2. Calabria CW, Dice J. Aeroallergen sensitization rates in military children with rhinitis symptoms. Ann Allergy Asthma Immunol. 2007;99(2):161-9.
  3. Gómez Castillo CA, Martínez Cairo Cueto S. [Diagnostic use of enzymatic RAST skin tests and determination of eosinophils in nasal mucosa in allergic rhinitis]. Rev Alerg Mex. 1998;45(6):150-8.
  4. Ortega EV, Vázquez MI, Tapia JG, Feria AJ. [Most common allergens in allergic patients admitted into a third-level hospital]. Rev Alerg Mex. 2004;51(4):145-50.
  5. Stern DA, Lohman IC, Wright AL, Taussig LM, Martinez FD, Halonen M. Dynamic changes in sensitization to specific aeroallergens in children raised in a desert environment. Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology. 2004;34(10):1563-669.
  6. Stokes JR, Hartel R, Ford LB, Casale TB. Cannabis (hemp) positive skin tests and respiratory symptoms. Ann Allergy Asthma Immunol. 2000;85(3):238-40.
  7. Kosrirukvongs P, Vichyanond P, Wongsawad W. Vernal keratoconjunctivitis in Thailand. Asian Pacific journal of allergy and immunology. 2003;21(1):25-30.