Taylor, a 29-year-old woman with a history of intermittent hives, visits her healthcare provider after developing abdominal cramps followed by hives five hours after eating a cheeseburger.
The most recent episode occurred at the emergency department where she works. Her coworkers noticed that her face and upper chest turned “beet red,” before she began coughing and wheezing. She was treated with albuterol nebulization, solumedrol, and epinephrine, which resolved her symptoms.
Her healthcare provider conducts a full clinical history and physical examination and decided on diagnostic testing that included the alpha-Gal component.
Taylor keeps waking up in the middle of the night covered in hives. What did specific IgE blood testing reveal?
These results together with this patient's case history and symptoms, help confirm the diagnosis.
| Test | Type | Taylor's Results |
| alpha-Gal | Allergen Component | 10.40 |
| Beef | Whole Allergen | 5.50 |
| Lamb | Whole Allergen | 4.60 |
| Pork | Whole Allergen | 5.10 |
| Total IgE | 28.60 |
Taylor has elevated levels of specific IgE to alpha-Gal, along with elevated beef, lamb and pork specific IgE indicating alpha-Gal syndrome.1
HEALTHCARE PROVIDER MANAGEMENT PLAN
FOLLOW UP
The people, places, and events depicted in these case studies and photographs do not represent actual patients, nor are they affiliated in any way with Thermo Fisher Scientific.
Before you proceed ...
We noticed you may be visiting a version of our website that doesn’t match your current location. Would you like to view content based on your region?
Talk with us
Interested in utilizing our solutions in your laboratory or healthcare practice?
This form is dedicated to healthcare and laboratory professionals who work within medical laboratories, private practices, health systems, and the like.
By clicking "continue with form" you are confirming that you work within a healthcare or laboratory space.
Not a lab professional or clinician but interested in diagnostic testing for allergies?
Visit Allergy InsiderChoose your preferred language
Please note: By selecting a different language, you are choosing to view another site. Product availability and indications may vary from what is approved in your region.