Abigail, a 19-year-old woman, visits her healthcare provider for guidance on her chronic gastrointestinal (GI) symptoms associated with eating meat.*
Her healthcare provider conducts a full clinical history and physical examination and decides to evaluate specific IgE sensitizations using both whole allergens and allergen components including alpha-Gal.
Abigail has chronic GI issues. What did specific IgE blood testing reveal?
These results together with this patient's case history and symptoms, help confirm the diagnosis.
| Test | Type | Abigail's Results |
| alpha-Gal | Allergen Component | 0.13 |
| Cat Dander | Whole Allergen | >100.00 |
| Cat Serum Albumin Fel d 2 | Allergen Component | 29.50 |
| Pork Serum Albumin | Allergen Component | 18.60 |
| Total IgE | 1406.00 |
Abigail has very low IgE levels to alpha-Gal, but has significant/high IgE to cat dander, cat serum albumin (Fel d 2), and pork serum albumin. Her GI symptoms could be explained by her sensitization to albumin, a protein that's found in both cat dander and in pork.1,2
HEALTHCARE PROVIDER MANAGEMENT PLAN
FOLLOW UP
The people, places, and events depicted in these case studies and photographs do not represent actual patients, nor are they affiliated in any way with Thermo Fisher Scientific.
*This case study was adapted from Platts-Mills T, Li R, Keshavarz B, Smith A, Wilson J. Diagnosis and Management of Patients with the α-Gal Syndrome. Journal of Allergy and Clinical Immunology: In Practice. 2020;8(1):15-23.e1.