Contact Us
Hidden Button for comparing instuments
Phadia 5000+ series Made to maximize
Phadia 500 instrument
Introduction to Phadia 5000+ series
Compare Instruments
These products may not be cleared for use in your country. Please contact your sales representative for information about specific product availability.
Sophisticated and powerful, the Phadia™ 5000+ series* provides the highest throughput of all our laboratory systems.
With four parallel processing lines, it doubles the capacity of the Phadia 2500+ series of instruments** and provides even greater flexibility. Our flagship solution is for extremely high throughput laboratories that deliver 25,000 to 45,000 results per week.
The three configuration options give you flexibility to acquire the most appropriate instrument to fit your laboratory needs, whether for ImmunoCAP™ only (Phadia 5000), or for both ImmunoCAP and EliA™ (Phadia 5000E and Phadia 5000E+E). This offers your laboratory the ability to customize a high-throughput automated solution that aligns with your testing needs and schedules. 
Thermo Fisher Phadia 2016. Photo: Markus Marcetic
Efficiency
  • Max throughput up to 960 ImmunoCAP results/hour or up to 300 EliA results/hour.
  • Continuous random access.
  • Refrigerated on-board reagent storage and 28-day stored calibration curves.
  • Onboard dilution (EliA).
  • Network multiple Phadia instruments for clustered*** setup for efficient handling.
Flexibility
  • Large allergen test menu covering more than 500 whole allergens and mixes and 100 allergen components.
  • Broad autoimmunity menu covering clinically relevant autoimmunity markers for evaluation of more than 50 of the most common autoimmune disease markers.
  • User-configurable reflex testing functionality.
  • LIS connectivity via PhadiaTM Prime software.
Confidence
  • Excellent consistency over time and between countries, systems, laboratories, and users.
  • Support and troubleshooting enhanced by the Phadia LabCommunity connectivity.
  • Quality Club: One of the largest global quality assessment programs.
*The Phadia 5000+ series comprises the Phadia 5000, Phadia 5000E, and Phadia 5000E+E instruments.
** The Phadia 2500+ series comprises the Phadia 2500, Phadia 2500E, and Phadia 2500EE instruments.
*** Cluster setup allows several Phadia instruments networked together to be operated with one Phadia Prime computer.
Phadia 5000+ series capability
Phadia 5000 testing instrument walk away time diagram
Explore the complete line of Phadia Laboratory Systems.
View all systems
See all tests that can be run on the Phadia 5000+ series.
View tests
View all available tests and instruments.
See Catalog