Search
Whether you are leveraging stem cells for basic research, drug discovery, or future therapeutic applications, we offer an extensive range of Gibco media, supplements, and reagents for all cell types to meet your stem cell culture needs. Manufactured with superior quality standards, our stem cell culture products offer consistency and accuracy to get the most from your experiments.
Pluripotent stem cell (PSC) research requires careful attention to culture conditions. From feeder-based media to feeder-free media to media for stem cell therapy research, Gibco products enable you to culture with confidence.
Gibco media and supplements are the most widely cited reagents for use in neural stem cell culture. Specifically formulated for serum-free growth and expansion of human neural stem cells, choose between B-27, N-2, StemPro Neural supplements, Neurobasal Medium, and more.
We offer an extensive selection of specialty products optimized for mesenchymal stem cell populations, many of which are free of animal-derived components and backed by our Gibco Cell Therapy Systems (CTS) brand. These media are designed to minimize adaptation time, maximize cell performance, and help meet regulatory requirements.
Hematopoietic stem cells expressing CD34 (CD34+ cells) are defined by their ability to replenish all blood cell types and to self-renew.Gibco StemPro-34 SFM is a serum-free medium specifically formulated to support the culture of these stem cells, and offers a superior expansion and differentiation of HSCs isolated from bone marrow, peripheral blood, or neonatal cord blood over traditional culture systems.
As you move from basic cell therapy research to the clinic, high-quality products and proper documentation are essential to getting it right the first time. Cell Therapy Systems (CTS) products help minimize the risk of contamination and variability in your research and provide all the required documentation for regulatory review—making them the superior choice as you transition from the bench to the clinic.
For Research Use Only. Not for use in diagnostic procedures.