Try new Gibco Insulin-Transferrin-Selenium Select supplements, an animal-origin free formulation. Available in ITS-G & ITS-X. Learn more >
Essential 8™ Flex Medium Kit
Gibco™

Essential 8™ Flex Medium Kit

Essential 8 Flex Medium is a xeno-free and feeder-free medium that supports the culture of pluripotent stem cells (PSCs) with a flexible feeding schedule.
Have Questions?
카탈로그 번호수량
A28585011 Kit
카탈로그 번호 A2858501
제품 가격(KRW)
549,000
온라인 행사
Ends: 31-Dec-2026
609,000
할인액 60,000 (10%)
Each
수량:
1 Kit
제품 가격(KRW)
549,000
온라인 행사
Ends: 31-Dec-2026
609,000
할인액 60,000 (10%)
Each

Essential 8 Flex Medium is a xeno-free and feeder-free medium that supports the culture of embryonic stem cells (ESCs) and induced pluripotent stem cells (iPSCs) without the need for weekend feeding. It is provided in a convenient two-component kit (500 mL basal medium and 10 mL supplement).

Essential 8 Flex provides all of the benefits of Essential 8 Medium: minimized variability of components, reliable and robust cultures with a xeno-free, cGMP produced medium, along with the added benefit of flexibility. Maintenance of standard PSCs no longer requires being in the lab seven days a week.

Flexibility

With Essential 8 Flex Medium, pluripotent stem cells (PSCs) need only be fed on the day of splitting and the day after. If the cells will be left without feeding for more than 48 hours, such as over a weekend, then simply double the feed volume on Friday.

Reliable and robust cultures

Essential 8 Flex Medium has been shown to support PSC growth for >30 passages without any signs of karyotypic abnormalities and to maintain the ability of iPSCs to differentiate into all three germ line lineages.

Cost effective

Essential 8 Flex Medium is provided in a convenient two-component kit (500 mL basal medium and 10 mL supplement), and when used with vitronectin (VTN-N), provides a cost effective, defined system for feeder-free culture of human pluripotent stem cells.

The Essential 8 Flex Medium formulation is based upon that published for Essential 8 Medium by Chen, et al., and allows for control and flexibility of PSC culture medium, without a negative impact to the health of the stem cells.

For Research Use Only. Not for use in diagnostic procedures.
사양
세포주Embryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs)
세포 유형Embryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs)
농도Essential 8 Flex Basal Medium (1X), Essential 8 Flex Supplement (50X)
제조 품질cGMP for medical devices, 21 CFR Part 820 and ISO 13485
제품라인Essential 8
제품 유형Stem Cell Media
수량1 Kit
유통 기한12 Months
배송 조건Supplement - dry ice, Basal - ambient
분류Xeno-free
Culture TypeAdherent Cell Culture, Feeder-free Stem Cell Culture (Human, iPS - Induced Pluripotent Stem, Embryonic), Stem Cell (Human, iPS - Induced Pluripotent Stem, Embryonic)
형태Liquid
Serum LevelSerum-free
멸균Sterile-filtered
Sterilization MethodSterile-filtered
첨가제 포함Phenol Red
Unit SizeEach
구성 및 보관
• 500 mL Basal Medium (store at 2–8°C and protect from light)
• 10 mL Supplement (store at -5 to -20°C and protect from light)

자주 묻는 질문(FAQ)

Do you still recommend using Gibco Vitronectin as the matrix for Gibco Essential 8 Flex Medium?

Both Gibco Vitronectin (VTN-N) Recombinant Human Protein, Truncated (Cat. No. A400457 and A400458) and Gibco Geltrex Flex LDEV-Free hESC-qualified Reduced Growth Factor Basement Membrane Matrix (Cat. No. A4000046801, A4000046802, or A4000046803) have been successfully used with Essential 8 Flex Medium.

Can PSCs transitioned over into Essential 8 Medium on recombinant human laminin-521 (rhLaminin-521) from feeder-dependent cultures, then be transferred onto vitronectin (VTN-N)?

Yes. Following 2 passages on the rhLaminin-521 matrix, Versene or EDTA passaging should be used to subculture PSCs.

What are the main differences to be taken into consideration when culturing cells in Gibco Essential 8 Medium on Gibco Vitronectin (VTN-N) compared to other feeder-free systems?

Here are three major differences to be taken into consideration when culturing cells in Gibco Essential 8 Medium on Gibco Vitronectin (VTN-N) compared to other feeder-free systems:
- Cells should be typically passaged ~24 hours sooner than they would be with other feeder-free media.
- Passaging should take place when cells are at ~85% confluency. If cells are passaged when they are more than 85% confluent, the health of the cells and final cell yield may be compromised.
- Cells must be passaged in EDTA. Collagenase and dispase are not recommended.

Can pluripotent stem cells (PSCs) previously cultured in other media and on other substrates be cultured in Gibco Essential 8 Medium and on Gibco Vitronectin?

Cells cultured in other feeder-free media systems, such as mTeSR Medium with Matrigel Basement Membrane Matrix, or StemPro hESC SFM with Geltrex Matrix, can be successfully cultured in Gibco Essential 8 Medium and VTN-N. In addition, PSCs grown on feeders with KnockOut SR have also been shown to be successfully cultured in Gibco Essential 8 Medium on VTN-N. However, when changing media systems, cells must be passaged either manually, or with EDTA prior to culturing in Gibco Essential 8 Medium on VTN-N.

Can I use enzymes such as dispase and collagenase for passaging cells cultured in Gibco Essential 8 Medium and on VTN-N?

Enzymes such as dispase and collagenase do not work well with cells cultured in Gibco Essential 8 Medium on VTN-N. Use of these enzymes for passaging cells results in compromised viability and attachment.

인용 및 참조 문헌 (1)

인용 및 참조 문헌
Abstract
Human-iPSC-Derived Cardiac Stromal Cells Enhance Maturation in 3D Cardiac Microtissues and Reveal Non-cardiomyocyte Contributions to Heart Disease.
Authors:Giacomelli E, Meraviglia V, Campostrini G, Cochrane A, Cao X, van Helden RWJ, Krotenberg Garcia A, Mircea M, Kostidis S, Davis RP, van Meer BJ, Jost CR, Koster AJ, Mei H, Míguez DG, Mulder AA, Ledesma-Terrón M, Pompilio G, Sala L, Salvatori DCF, Slieker RC, Sommariva E, de Vries AAF, Giera M, Semrau S, Tertoolen LGJ, Orlova VV, Bellin M, Mummery CL
Journal:Cell Stem Cell
PubMed ID:32459996
'Cardiomyocytes (CMs) from human induced pluripotent stem cells (hiPSCs) are functionally immature, but this is improved by incorporation into engineered tissues or forced contraction. Here, we showed that tri-cellular combinations of hiPSC-derived CMs, cardiac fibroblasts (CFs), and cardiac endothelial cells also enhance maturation in easily constructed, scaffold-free, three-dimensional microtissues (MTs). ... More