CTS™ Versene™ Solution

CTS™ Versene™ Solution

CTS Versene Solution is a cGMP-manufactured EDTA solution for use as a gentle non-enzymatic cell dissociation reagent.
Have Questions?
Catalog NumberQuantity
A4239101100 mL
Catalog number A4239101
Price (USD)
44.54
Each
Quantity:
100 mL
Price (USD)
44.54
Each

CTS Versene Solution is a cGMP-manufactured EDTA solution for use as a gentle non-enzymatic cell dissociation reagent. It is formulated as 0.2 g EDTA(Na4) per liter of phosphate buffered saline (PBS) (0.48 mM). It reduces the burden in qualifying reagents when transitioning from the research bench to the clinic.

Maintain viability over multiple passages

PSCs passaged with this cGMP manufactured EDTA dissociation reagent maintain viability.

Designed with transition in mind

CTS Versene Solution has the same formulation as RUO Versene Solution but with additional characterization and quality control testing. CTS Versene Solution is manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing site is ISO 13485-certified. Full documentation traceability is available for CTS Versene Solution.

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Specifications
Cell TypePluripotent Stem Cells (PSCs)
Concentration1X
For Use With (Application)Cell and Gene Therapy Research, Development, and Manufacturing
FormatBottle
Manufacturing QualityISO 13485, MDSAP, FDA-registered, 21 CFR 820
Quantity100 mL
Shelf Life18 months from date of manufacture
Shipping ConditionRoom Temperature
ClassificationAnimal Origin-free
FormLiquid
Product TypeCell Culture Dissociation Reagent
SterilitySterile-filtered
Unit SizeEach
Contents & Storage
Store at 2–8°C and protect from light

Frequently asked questions (FAQs)

What is CTS?

The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.