PhadiaTM Laboratory Systems are designed to help your laboratory deliver successful test results, quickly and efficiently. With high sensitivity and specificity, these highly automated systems use fluorescence enzyme immunoassay (FEIA) technology to offer the superior ability to detect and quantitatively determine specific antibodies in blood.
By processing samples from test requests to final results with minimal interventions, Phadia Laboratory Systems reduce hands-on time and improve your workflow:
Easier audits and set-up with barcode identification of all reagents
User efficiency and minimized errors with mainframe worklist downloads, barcode sample identification, and on-board dilution
Faster loading times with on-board storage of all reagents
Ability to run ImmunoCAPTM and EliATM assays on the same system*
*The Phadia 1000 instrument processes ImmunoCAP assay exclusively
Designed to simplify and optimize daily operation
Phadia Prime software optimizes processing of ImmunoCAP and EliA assays on Phadia Laboratory Systems to aid in the diagnosis of allergic and autoimmune diseases. Innovative features of Phadia Prime that help your laboratory meet today’s workflow demands include:
Intuitive, real-time management of reagent inventory
Extended audit trail
report
Enterprise Resource Planning (ERP) integration
Two-way LIS channel communication and dual LIS channel capabilities
Test prioritization by adult/pediatric samples
Integrated Quantity Not Sufficient (QNS) handling
Customizable reflex testing capabilities to support diagnostic pathways
This state-of-the-art data management software allows efficient management of multiple Phadia instruments. Phadia Prime software, an integral part of Phadia Laboratory Systems, continuously helps improve productivity in clinical laboratories.