+
For Patients & Caregivers
For Healthcare Professionals
Welcome! Click here for Patient or Healthcare Professional content
Are you a healthcare professional?
The information in this website is intended only for healthcare professionals. By entering this site, you are confirming that you are a healthcare professional.
Are you a laboratory professional?
The information in this website is intended only for laboratory professionals. By entering this site, you are confirming that you are a laboratory professional.
Phadia LabCommunity
MAXIMIZE THE PRODUCTIVITY IN YOUR LABORATORY
Developed to provide real-time support
For a laboratory like yours, maintaining the integrity of your data is a way to ensure your productivity. PhadiaTM LabCommunity software monitors your Phadia Laboratory System to help minimize the risk of losing valuable data and time. Data integrity and security is ensured by thorough security measures, such as secure server connections, removal of patient data, encrypted data transmission, user permission control and activity logging. The end result is the potential to maximize your laboratory’s instrument up time while reducing the cost of dealing with corrupted data.
Features of Phadia LabCommunity
  • Encrypted remote data connection to facilitate service and support
  • Automatic removal of sensitive data before data collection
  • Remote assistance
  • Automatic reporting of Quality Club results
Lab Community
How Phadia LabCommunity Works
1. Your Phadia Laboratory Systems
  • Connection to Phadia LabCommunity is easily activated through Phadia Prime settings
  • Automatic transition of system status messages
  • Firewall set-up for secure connections
2. Secure Data Transmission
  • Your laboratory’s computer communicates with our server over an encrypted, Secure connection using https and Secure Sockets Layer (SSL)
  • Supports multiple network configurations
3. Our Server
  • Firewall protection and SSL server-side certificates
  • Our server is protected by restrictions in access rights
4. Our Service and Support Specialists
  • Accessible only to certified personnel