Thermo Scientific Chemicals

Sodium dihydrogen phosphate monohydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 010870.22
100 g, Each
Thermo Scientific Chemicals

Sodium dihydrogen phosphate monohydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 010870.22
100 g, Each
Quantity
Catalog number: 010870.22
also known as 010870-22
Price (USD)
Price: 443.00
Online price: 376.65
Your price:
Quantity
-

Chemical Identifiers

CAS
10049-21-5
IUPAC Name
sodium phosphoric acid hydrate
Molecular Formula
H5NaO5P
InChI Key
BBMHARZCALWXSL-UHFFFAOYSA-N
SMILES
O.[Na+].OP(O)(O)=O
Total Metal Impurities
0.002% max.

Description

Sodium dihydrogen phosphate monohydrate is used as a food additive. It finds use as mordant in dyeing. It is also used in the manufacture of enamels, chemicals, pharmaceuticals, ceramics, detergents, and boiler compounds. In addition, it is used as a fireproofing agent, thickening agent, emulsifier, and as a reagent and buffer in analytical chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text