Thermo Scientific Chemicals

Sodium fluoride, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 011003.22
100 g, Each
Thermo Scientific Chemicals

Sodium fluoride, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 011003.22
100 g, Each
Quantity

Chemical Identifiers

CAS
7681-49-4
IUPAC Name
sodium fluoride
Molecular Formula
FNa
InChI Key
PUZPDOWCWNUUKD-UHFFFAOYSA-M
SMILES
[F-].[Na+]
Appearance (Color)
White
Assay (special)
?2.5 % of Theoretical value (Na)
Total Metal Impurities
≤0.01% (100 ppm)
Form
Powder

Description

It is used as a steel degreasing agent and in tin electroplating, flux, heat treating salt compositions, fluoridation of drinking water, removing HF from exhaust systems to reduce air pollution, and frosting glasses. It serves as a source of fluoride ion in several applications including its use in synthetic chemistry, for example in Finkelstein reaction for preparing fluoro compounds, desilylation in sensitive organic compounds, metallurgy, and reaction with electrophilic chlorides. It has medical and other applications including complexing of magnesium ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text