Thermo Scientific Chemicals

Cerium(IV) sulfate hydrate, REacton™, 99% (REO), Thermo Scientific Chemicals

Catalog number: 011323.36
500 g, Each
Thermo Scientific Chemicals

Cerium(IV) sulfate hydrate, REacton™, 99% (REO), Thermo Scientific Chemicals

Catalog number: 011323.36
500 g, Each
Quantity
Catalog number: 011323.36
also known as 011323-36
Price (USD)
Price: 434.00
Online price: 375.65
Your price:
Quantity
-

Chemical Identifiers

CAS
95838-16-7
IUPAC Name
λ⁴-cerium(4+) disulfate
Molecular Formula
CeO8S2
InChI Key
VZDYWEUILIUIDF-UHFFFAOYSA-J
SMILES
[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Total Rare Earth Oxide Impurities
1% max.

Description

Cerium(IV) sulfate hydrate acts as oxidizing agent which is used for the generation of o-benzoquinone from catechol. It promotes the formation of radicals from dialkyl malonates, which undergo substitution reactions with furans and thiophenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text