Thermo Scientific Chemicals

Sodium perrhenate, 99+% (metals basis), Re 68%, Thermo Scientific Chemicals

Catalog number: 011412.09
10 g, Each
Thermo Scientific Chemicals

Sodium perrhenate, 99+% (metals basis), Re 68%, Thermo Scientific Chemicals

Catalog number: 011412.09
10 g, Each
Quantity
Catalog number: 011412.09
also known as 011412-09
Price (USD)
Price: 366.00
Online price: 311.65
Your price:
Quantity
-

Chemical Identifiers

CAS
13472-33-8
IUPAC Name
sodium rheniumoylolate
Molecular Formula
NaO4Re
InChI Key
KMISVFTVBOPTAI-UHFFFAOYSA-N
SMILES
[Na+].[O-][Re](=O)(=O)=O
Total Metal Impurities
1% max., Re 68%

Description

Sodium perrhenate is used as a precursor to other rhenium compounds. Further, it reacts with sodium in ethanol to prepare nonahydridorhenate and with polysulfide solutions to get tetrathiorhenate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text