Thermo Scientific Chemicals

Silver cyanide, 99%, Thermo Scientific Chemicals

Catalog number: 011424.22
100 g, Each
Thermo Scientific Chemicals

Silver cyanide, 99%, Thermo Scientific Chemicals

Catalog number: 011424.22
100 g, Each
Quantity
Catalog number: 011424.22
also known as 011424-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
506-64-9
IUPAC Name
silver(1+) iminomethanide
Molecular Formula
CAgN
InChI Key
LFAGQMCIGQNPJG-UHFFFAOYSA-N
SMILES
[Ag+].[C-]#N
Assay (unspecified)
98.6% min

Description

Silver cyanide is used in silver-plating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text