Thermo Scientific Chemicals

Vanadium(IV) fluoride, 95%, Thermo Scientific Chemicals

Catalog number: 011543.06
5 g, Each
Thermo Scientific Chemicals

Vanadium(IV) fluoride, 95%, Thermo Scientific Chemicals

Catalog number: 011543.06
5 g, Each
Quantity
Catalog number: 011543.06
also known as 011543-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
10049-16-8
IUPAC Name
vanadium pentafluoride
Molecular Formula
F5V
InChI Key
LYTSFXQJUMZNFC-UHFFFAOYSA-N
SMILES
[F].[F].[F].[F].[F].[V]
Assay (unspecified)
94.1% min

Description

Vanadium(IV) fluoride is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text