Thermo Scientific Chemicals

Manganese(II) perchlorate hexahydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 012309.22
100 g, Each
Thermo Scientific Chemicals

Manganese(II) perchlorate hexahydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 012309.22
100 g, Each
Quantity
Catalog number: 012309.22
also known as 012309-22
Price (USD)
Price: 48.00
Online price: 41.65
Your price:
Quantity
-

Chemical Identifiers

CAS
15364-94-0
IUPAC Name
manganese(2+) hexahydrate diperchlorate
Molecular Formula
Cl2H12MnO14
InChI Key
AIFGVOMWPFMOCN-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.[Mn++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Assay (unspecified)
Mg 14.9-15.9%

Description

It used in rocket fuel. This is one of the most proven materials for etching of liquid crystal displays and critical electronics applications as well as ore extraction. It is also used for electropolishing/etching of aluminum, molybdenum, and other metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text