Indium(III) 2,4-pentanedionate, 98%
Indium(III) 2,4-pentanedionate, 98%
Indium(III) 2,4-pentanedionate, 98%
Thermo Scientific Chemicals

Indium(III) 2,4-pentanedionate, 98%

Used in catalysts | CAS: 14405-45-9 | C15H21InO6 | 412.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 013794.03
also known as 013794-03
Price (USD)
35.80
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
35.80
Each
Chemical Identifiers
CAS14405-45-9
IUPAC Nameindium(3+) tris(2,4-dioxopentan-3-ide)
Molecular FormulaC15H21InO6
InChI KeyCMGVNFXMZKDHDG-UHFFFAOYSA-N
SMILES[In+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)<0.30% Volatiles
Insoluble material<0.30%
FormCrystalline powder
Used in catalysts.Indium(III) 2,4-pentanedionate is used as a precursor for the preparation of indium(III)oxide. It acts as a catalyst in the synthesis of oxindole derivatives. It finds application in gas sensors. Further, it is used in the synthesis of indium tin oxide nanocrystals. In addition to this, it is involved in the preparation of thin-film solar cells by atomic layer chemical vapor deposition with hydrogen sulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only