Tantalum plate, 6.35mm (0.25in) thick, 99.9% (metals basis)
Tantalum plate, 6.35mm (0.25in) thick, 99.9% (metals basis)
Tantalum plate, 6.35mm (0.25in) thick, 99.9% (metals basis)
Thermo Scientific Chemicals

Tantalum plate, 6.35mm (0.25in) thick, 99.9% (metals basis)

CAS: 7440-25-7 | Ta | 180.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 014181.FF
also known as 014181-FF
Price (USD)
522.65
Online Exclusive
580.00
Save 57.35 (10%)
Each
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)
522.65
Online Exclusive
580.00
Save 57.35 (10%)
Each
Specifications
Assay Percent Notes(metals basis)
CAS7440-25-7
ChEBICHEBI:33348
Chemical Name or MaterialTantalum plate
EINECS Number231-135-5
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only