Thermo Scientific Chemicals

Manganese(IV) oxide, activated, tech., Mn 58% min, Thermo Scientific Chemicals

Catalog number: 014340.09
10 g, Each
Thermo Scientific Chemicals

Manganese(IV) oxide, activated, tech., Mn 58% min, Thermo Scientific Chemicals

Catalog number: 014340.09
10 g, Each
Quantity
Catalog number: 014340.09
also known as 014340-09
Price (USD)
Price: 30.70
Online price: 26.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1313-13-9
IUPAC Name
dioxomanganese
Molecular Formula
MnO2
InChI Key
NUJOXMJBOLGQSY-UHFFFAOYSA-N
SMILES
O=[Mn]=O
Assay (unspecified)
58% Mn min
Particle Size
5-13 Microns
Assay (unspecified)
16% available oxygen min
Impurity content
0.7-1.3% NaOH
Residual water
2% max

Description

As an oxidizing agent, source of metallic manganese, decolorizing glass, painting porcelain, as an analytical reagentManganese(IV) oxide is used as an oxidizing agent in organic synthesis such as oxidation of allylic/benzylic alcohols, as a textile dye, as a reducing agent, and as a component of dry cell batteries such as zinc-carbon battery and alkaline battery. It is also used in making pigments for glasses and ceramics, and drier for paints. Further it is used in the manufacture of manganese steel and several manganese derivatives including potassium permanganate, a powerful versatile oxidant. Catalytic hydration of nitriles to amides by flowing through manganese dioxide has been reported. Manganese dioxide supported on inorganic oxide can be used for oxidation of methylamine through CWAO (Catalytic Wet Air Oxidation). It has high potential as highly efficient and robust material for water oxidation reaction (WORs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • As an oxidizing agent, source of metallic manganese, decolorizing glass, painting porcelain, as an analytical reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text