Thermo Scientific Chemicals

Thymol Blue, ACS

Catalog number: 016272.04
2 g, Each
Thermo Scientific Chemicals

Thymol Blue, ACS

Catalog number: 016272.04
2 g, Each
Quantity
Catalog number: 016272.04
also known as 016272-04
Price (USD)
Price: 21.70
Online price: 18.65
Your price:
Quantity
-

Chemical Identifiers

CAS
76-61-9
IUPAC Name
3,3-bis[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular Formula
C27H30O5S
InChI Key
PRZSXZWFJHEZBJ-UHFFFAOYSA-N
SMILES
CC(C)C1=CC(=C(C)C=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C(C)C)=C(O)C=C1C
Comment
Clarity of solution P.T.
Comment
Visual transition interval (acid range) pH 1.2 (pink) to 2.8 (yellow)
Comment
Visual transition range (alkaline range) pH 8.0 (yellow) to 9.2 (blue)

Description

pH indicator: 1.2 (pink) to 2.8 (yellow) to 9.6 (blue). Thymol Blue is useful as a pH indicator. It is a component of universal indicator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text