Thermo Scientific Chemicals

2,6-Dimethyl-5-heptenal, stabilized, Thermo Scientific Chemicals

Catalog number: 017773.30
250 g, Each
Thermo Scientific Chemicals

2,6-Dimethyl-5-heptenal, stabilized, Thermo Scientific Chemicals

Catalog number: 017773.30
250 g, Each
Quantity
Catalog number: 017773.30
also known as 017773-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
106-72-9
IUPAC Name
2,6-dimethylhept-5-enal
Molecular Formula
C9H16O
InChI Key
YGFGZTXGYTUXBA-UHFFFAOYNA-N
SMILES
CC(CCC=C(C)C)C=O
Assay (Titration)
≥85.0%
Comment
Contains 0.1 to 1.5% dl-alpha-Tocopherol
Refractive Index
1.4420 to 1.4470
Specific Gravity
0.848 - 0.854
Form
Liquid

Description

It is used or fragrances designed to have a special fruity note tending towards melon, cucumber and pumpkin. It is mainly used perfuming agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text