Sodium metatungstate monohydrate
Thermo Scientific Chemicals

Sodium metatungstate monohydrate

CAS: 314075-43-9 | Na6O39W12·H2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
4 x 250 g
Catalog number 022534.56
also known as 022534-56
Price (USD)
-
Quantity:
4 x 250 g
Request bulk or custom format
Chemical Identifiers
CAS314075-43-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White or pale green
FormCrystals or powder or crystalline powder or granules
Assay (unspecified)≥87.0% WO₃
Sodium metatungstate monohydrate plays a vital role in sink and swim analysis. It is a derivative of sodium tungstate, which is a potential therapy for obesity. It acts as an inhibitor, which inhibits nucleoside-triphospahte- diphosphohydrolases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only