Crystal Violet, ACS, 90+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Crystal Violet, ACS, 90+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number 022866.22
also known as 022866-22
Price (USD)/ Each
82.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
97.10 
Save 14.45 (15%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
82.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
97.10 
Save 14.45 (15%)
Add to cart
Crystal Violet, ACS, 90+%, Thermo Scientific Chemicals
Catalog number022866.22
Price (USD)/ Each
82.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
97.10 
Save 14.45 (15%)
-
Add to cart
Chemical Identifiers
CAS548-62-9
IUPAC Nametris[4-(dimethylamino)phenyl]methylium chloride
Molecular FormulaC25H30ClN3
InChI KeyZXJXZNDDNMQXFV-UHFFFAOYSA-M
SMILES[Cl-].CN(C)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark green to metallic green
Assay from Supplier's CofA≥90.0%
FormPowder or crystals or crystalline powder
CommentSensitivity as indicator P.T.
CommentAbsorbance characteristics P.T.
View more
Crystal Violet is used as an acid-base indicator, an alcohol denaturant, a biological stain, a textile dye and an indicator for copper salts. It is a component of gram staining in microorganisms and cell lines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only