1,2-Propanediol, ACS, 99.5% min
1,2-Propanediol, ACS, 99.5% min
1,2-Propanediol, ACS, 99.5% min
Thermo Scientific Chemicals

1,2-Propanediol, ACS, 99.5% min

As a substitute for ethylene glycol and glycerol. Good solvent for resins. Dissolves many essential oils, but immiscible with fixed oils. | CAS: 57-55-6 | C3H8O2 | 76.095 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
4 x 1 L
Catalog number 030948.M1
also known as 030948-M1
Price (USD)
261.00
Each
Quantity:
4 x 1 L
Request bulk or custom format
Price (USD)
261.00
Each
Chemical Identifiers
CAS57-55-6
IUPAC Namepropane-1,2-diol
Molecular FormulaC3H8O2
InChI KeyDNIAPMSPPWPWGF-UHFFFAOYNA-N
SMILESCC(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤0.2%
FormViscous liquid
Impurity contentCl: ≤1ppm
Color (APHA)≤10
Assay (GC)≥99.5% ( corrected for water content)
View more
1,2-Propanediol, ACS is used as a substitute for ethylene glycol and glycerol. It is a good solvent for resins. It dissolves many essential oils but is immiscible with fixed oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only