Thermo Scientific Chemicals

Iron(III) bromide, anhydrous, 98+%, Thermo Scientific Chemicals

Catalog number: 031125.22
100 g, Each
Thermo Scientific Chemicals

Iron(III) bromide, anhydrous, 98+%, Thermo Scientific Chemicals

Catalog number: 031125.22
100 g, Each
Quantity
Catalog number: 031125.22
also known as 031125-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10031-26-2
IUPAC Name
iron(3+) tribromide
Molecular Formula
Br3Fe
InChI Key
FEONEKOZSGPOFN-UHFFFAOYSA-K
SMILES
[Fe+3].[Br-].[Br-].[Br-]
Appearance (Color)
Red to black
Form
Powder or crystals or crystalline powder
Assay (unspecified)
≥98.0%
Water Content (Karl Fischer Titration)
≤2%

Description

Iron(III) bromide is used as a Lewis acid catalyst in the halogenation of aromatic compounds. It is also used in the preparation of 2,5-dibromo-4-aryl-1,3-pentadiene by reacting with homoallenic bromohydrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text