Thermo Scientific Chemicals

Magnesium sulfate, anhydrous, 99.5% min, Thermo Scientific Chemicals

Catalog number: 033337.A3
2 kg, Each
Thermo Scientific Chemicals

Magnesium sulfate, anhydrous, 99.5% min, Thermo Scientific Chemicals

Catalog number: 033337.A3
2 kg, Each
Quantity
Catalog number: 033337.A3
also known as 033337-A3
Price (USD)
Price: 144.00
Online price: 122.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7487-88-9
IUPAC Name
magnesium(2+) sulfate
Molecular Formula
MgO4S
InChI Key
CSNNHWWHGAXBCP-UHFFFAOYSA-L
SMILES
[Mg++].[O-]S([O-])(=O)=O
Form
Powder
Loss on Drying
2% max
Trace Metals
0.001% max
Impurity content
0.003% Se max
Assay (unspecified)
99.5% min

Description

Magnesium sulfate is suitable for sensitive molecular biology applications. It used as a component of bath salts and beauty product. It is utilized by the athletes to soothe sore muscles and effective in the removal of splinters. It is commonly used as a desiccant in organic synthesis due to its affinity for water. It acts as a brewing salt in beer production to adjust the ion content of the brewing water as well as enhance enzyme action. It plays a vital role as an antidote for barium chloride poisoning. . It is useful in controlling seizures associated with epilepsy, glomerulonephritis, or hypothyroidism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text