Thermo Scientific Chemicals

Barium bromide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 035678.03
1 g, Each
Thermo Scientific Chemicals

Barium bromide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 035678.03
1 g, Each
Quantity
Catalog number: 035678.03
also known as 035678-03
Price (USD)
Quantity
-

Chemical Identifiers

CAS
10553-31-8
Residual water
< 100 ppm
Total Metal Impurities
0.002% max.
Particle Size
-10 Mesh
Appearance (Color)
White
Form
Beads

Description

Barium bromide is a precursor to chemicals used in photography and to other bromides. Barium bromide was used to purify radium in a process of fractional crystallization devised by Marie Curie. Since radium precipitates preferentially in a solution of barium bromide, the ratio of radium to barium in the precipitate would be higher than the ratio in the solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text