Thermo Scientific Chemicals

4-Amino-3-hydroxy-1-naphthalenesulfonic acid, ACS, 90+%, Thermo Scientific Chemicals

Catalog number: 036290.22
100 g, Each
Thermo Scientific Chemicals

4-Amino-3-hydroxy-1-naphthalenesulfonic acid, ACS, 90+%, Thermo Scientific Chemicals

Catalog number: 036290.22
100 g, Each
Quantity
Catalog number: 036290.22
also known as 036290-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
116-63-2
IUPAC Name
4-amino-3-hydroxynaphthalene-1-sulfonic acid
Molecular Formula
C10H9NO4S
InChI Key
RXCMFQDTWCCLBL-UHFFFAOYSA-N
SMILES
NC1=C2C=CC=CC2=C(C=C1O)S(O)(=O)=O
Solution Test
in sodium carbonate P.T.
Comment
Sensitivity to phosphate P.T.
Impurity content
SO₄ 0.2%
Residue on ignition
0.1%

Description

4-Amino-3-hydroxy-1-naphthalenesulfonic acid is used as a key starting material for the preparation of naphthalenesulfonic acid derivatives by reacting with palmitoyl chloride, which shows the activity to inhibit DNA polymerase and ribonuclease H Activities of HIV-1 reverse transcriptase. It is also used to prepare a copolyner with melamine, which is associated with graphene to prepare molecularly imprinted sensor and utilized for the determination of melatonin in human biological fluids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text