Thermo Scientific Chemicals

Sodium formate, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 036424.36
500 g, Each
Thermo Scientific Chemicals

Sodium formate, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 036424.36
500 g, Each
Quantity
Catalog number: 036424.36
also known as 036424-36
Price (USD)
Price: 64.70
Online price: 54.65
Your price:
Quantity
-

Chemical Identifiers

CAS
141-53-7
IUPAC Name
sodium formate
Molecular Formula
CHNaO2
InChI Key
HLBBKKJFGFRGMU-UHFFFAOYSA-M
SMILES
[Na+].[O-]C=O
Assay (unspecified)
99.0% HCO₂Na min
Impurity content
Cl 0.001%
Impurity content
Fe 5ppm
Impurity content
SO₄ 0.001%
Insoluble material
0.005%

Description

Reducing agent, buffering agent, complexing agent, Noble metal precipitantSodium formate is used in analytical chemistry to precipitate noble metals. It is used for tanning leather, for dyeing and printing fabrics. It acts as an electroplating agent, an acidulant in the textile industry, a reducing agent, a mordant, a complexing agent. Further, it is used in photographic fixing baths. In addition, it is used as a buffering agent for strong mineral acids and food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text