Antimony(III) isopropoxide, 99.9% (metals basis)
Antimony(III) isopropoxide, 99.9% (metals basis)
Antimony(III) isopropoxide, 99.9% (metals basis)
Thermo Scientific Chemicals

Antimony(III) isopropoxide, 99.9% (metals basis)

CAS: 18770-47-3 | C9H21O3Sb | 299.024 g/mol
製品番号(カタログ番号) 036589.06
または、製品番号036589-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
仕様
定量パーセント注記(metals basis)
CAS18770-47-3
化学物質名または材質Antimony(III) isopropoxide
Colorless
DOT情報Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only