Thermo Scientific Chemicals

Bromopentacarbonylmanganese(I), 98%, Thermo Scientific Chemicals

Catalog number: 039310.06
5 g, Each
Thermo Scientific Chemicals

Bromopentacarbonylmanganese(I), 98%, Thermo Scientific Chemicals

Catalog number: 039310.06
5 g, Each
Quantity
Catalog number: 039310.06
also known as 039310-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14516-54-2
IUPAC Name
pentakis(methanidylidyneoxidanium) manganese bromide
Molecular Formula
C5BrMnO5
InChI Key
LOCZKKJCQVRJPQ-UHFFFAOYSA-M
SMILES
[Mn].[Br-].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
Appearance (Color)
Orange to dark orange
Assay from Suppliers CofA
≥97.5%
Form
Crystals or powder or crystalline powder

Description

Bromopentacarbonylmanganese(I) is used in the formation of (eta6-arene)tricarbonylmanganese(I) by reacting with arene (arene= hexamethyl benzene, 1,2,4,5-tetramethyl benzene, mesitylene, p-xylene and toluene) in the presence silver salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text