Gold powder, spherical, APS 0.8-1.5 micron, 99.96+% (metals basis)
Gold powder, spherical, APS 0.8-1.5 micron, 99.96+% (metals basis)
Gold powder, spherical, APS 0.8-1.5 micron, 99.96+% (metals basis)
Thermo Scientific Chemicals

Gold powder, spherical, APS 0.8-1.5 micron, 99.96+% (metals basis)

Apparent density 40-70g/in3, Tap density: 4.0-9.0g/cm3 | CAS: 7440-57-5 | Au | 196.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number 039817.03
also known as 039817-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay from Supplier's CofA>99.96% (metals basis)
Assay from Supplier's CofAAg < 0.008%
Assay from Supplier's CofACu < 0.005%
Assay from Supplier's CofAFe < 0.005%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only