Thermo Scientific Chemicals

Nickel(II) carbonate (basic) hydrate, Ni 40% min, typically 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 040106.A1
1 kg, Each
Thermo Scientific Chemicals

Nickel(II) carbonate (basic) hydrate, Ni 40% min, typically 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 040106.A1
1 kg, Each
Quantity
Catalog number: 040106.A1
also known as 040106-A1
Price (USD)
Price: 124.00
Online price: 105.65
Your price:
Quantity
-

Chemical Identifiers

CAS
155775-31-8
IUPAC Name
trinickel(2+) tetrahydroxide carbonate
Molecular Formula
CH4Ni3O7
InChI Key
ATYNVYNPWGEMMI-UHFFFAOYSA-H
SMILES
[OH-].[OH-].[OH-].[OH-].[Ni++].[Ni++].[Ni++].[O-]C([O-])=O
Assay (unspecified)
Ni 40% min

Description

Nickel plating pH adjuster, catalysts, ceramic colors and glazes.Nickel(II) carbonate (basic) hydrate is used as a precursor to catalysts and in ceramic applications. It acts as an intermediate in the hydrometallurgical purification of nickel from its ores. It is also used in electroplating of nickel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text