Thermo Scientific Chemicals

Cobalt naphthenate, Co 6%, approximately 53% in mineral spirits, Thermo Scientific Chemicals

Catalog number: 040387.A3
2 kg, Each
Thermo Scientific Chemicals

Cobalt naphthenate, Co 6%, approximately 53% in mineral spirits, Thermo Scientific Chemicals

Catalog number: 040387.A3
2 kg, Each
Quantity
Catalog number: 040387.A3
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
61789-51-3
IUPAC Name
λ²-cobalt(2+) bis(3-(3-ethylcyclopentyl)propanoate)
Molecular Formula
C20H34CoO4
InChI Key
XSWKLHINRKWMTD-UHFFFAOYNA-L
SMILES
[Co++].CCC1CCC(CCC([O-])=O)C1.CCC1CCC(CCC([O-])=O)C1
Appearance (Color)
Purple
Elemental Analysis
Co: 5.5-6.5%
Form
Liquid

Description

Cobalt naphthenate is widely used in polyester resins and paint driers. It is used as a curing accelerator, cross linker catalyst for unsaturated polyester resins. It is also used in the production of adhesives, varnishes and waterproofing agents for textiles. Further, it is used as corrosion inhibitors, lubricants, and fuel additives. It plays an important role as oil drying agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text