Thermo Scientific Chemicals
Cerium(IV) 2-methoxyethoxide, 18-20% w/w in 2-methoxyethanol, Thermo Scientific Chemicals
Catalog number: 040471.22
100 g, Each
Thermo Scientific Chemicals
Cerium(IV) 2-methoxyethoxide, 18-20% w/w in 2-methoxyethanol, Thermo Scientific Chemicals
Catalog number: 040471.22
100 g, Each
Quantity
Catalog number: 040471.22
also known as 040471-22
Price (USD)
Quantity
-
Description
Hydrolysis of cerium 2-methoxyethoxide and subsequent supercritical drying or freeze-drying yielded a CeO 2 aerogel or cryogel with quite a large surface area above 300 m 2 /g.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov