Aluminum ethoxide
Aluminum ethoxide
Aluminum ethoxide
Thermo Scientific Chemicals

Aluminum ethoxide

CAS: 555-75-9 | C6H15AlO3-3 | 162.165 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 041336.30
also known as 041336-30
Price (USD)
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS555-75-9
IUPAC Namealuminium(3+) triethanolate
Molecular FormulaC6H15AlO3
InChI KeyJPUHCPXFQIXLMW-UHFFFAOYSA-N
SMILES[Al+3].CC[O-].CC[O-].CC[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder and chunk
Elemental AnalysisAl > 16.4%
Appearance (Color)White
Aluminum ethoxide is mainly used in industrial settings. It is used as a reducing agent for aldehyde, ketones, polymerization catalyst. Used in the sol-gel process preparation of high purity aluminum sesquioxide. It also used as a reducing reagent, for carbonyl compounds that restore to alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only